Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 149572869-149573110 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WWTR1
Entrez Gene ID
25937
Gene Name
WW domain containing transcription regulator 1
Gene Aliases
TAZ
UniGene
Hs.477921
Gene Chromosome Location
Chr.3: 149517235-149724783 on Build GRCh38
Transcripts
NM_001168278.1 NM_001168280.1 NM_015472.4 XM_017006121.1 XM_017006122.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4114357 c.514G>A p.V172I Chr.3: 149572918-149572918 on Build GRCh38
7397982 c.464C>G p.P155R Chr.3: 149572968-149572968 on Build GRCh38
6823643 c.483G>A p.Q161Q Chr.3: 149572949-149572949 on Build GRCh38
5863726 c.512C>T p.A171V Chr.3: 149572920-149572920 on Build GRCh38
3915112 c.504C>T p.L168L Chr.3: 149572928-149572928 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751244616 Chr.3: 149572905-149572905 on Build GRCh38
rs371622113 Chr.3: 149573025-149573025 on Build GRCh38
rs774219291 Chr.3: 149572950-149572950 on Build GRCh38
rs765590185 Chr.3: 149573015-149573015 on Build GRCh38
rs755578307 Chr.3: 149572930-149572930 on Build GRCh38
rs749185901 Chr.3: 149572935-149572935 on Build GRCh38
rs377048586 Chr.3: 149572918-149572918 on Build GRCh38
rs764139095 Chr.3: 149572994-149572994 on Build GRCh38
rs534445202 Chr.3: 149572958-149572958 on Build GRCh38
rs777900756 Chr.3: 149572931-149572931 on Build GRCh38
rs200341740 Chr.3: 149573047-149573047 on Build GRCh38
rs755502128 Chr.3: 149573017-149573020 on Build GRCh38
rs943035430 Chr.3: 149573058-149573058 on Build GRCh38
rs544086561 Chr.3: 149573035-149573035 on Build GRCh38
rs557309752 Chr.3: 149572962-149572962 on Build GRCh38
rs201045484 Chr.3: 149572991-149572991 on Build GRCh38
rs775570561 Chr.3: 149572959-149572959 on Build GRCh38
rs375892171 Chr.3: 149573041-149573041 on Build GRCh38
rs762268907 Chr.3: 149573006-149573006 on Build GRCh38
rs757669865 Chr.3: 149572892-149572892 on Build GRCh38
rs565403324 Chr.3: 149572904-149572904 on Build GRCh38
rs914741890 Chr.3: 149572927-149572927 on Build GRCh38
rs763217811 Chr.3: 149573030-149573030 on Build GRCh38
rs780584391 Chr.3: 149572922-149572922 on Build GRCh38
rs752060235 Chr.3: 149573012-149573012 on Build GRCh38
rs574280322 Chr.3: 149573081-149573081 on Build GRCh38
rs886119968 Chr.3: 149572980-149572980 on Build GRCh38
rs947633979 Chr.3: 149573004-149573004 on Build GRCh38
rs770878112 Chr.3: 149572936-149572936 on Build GRCh38
rs376585599 Chr.3: 149572956-149572956 on Build GRCh38
rs1013529773 Chr.3: 149572908-149572908 on Build GRCh38
rs777145948 Chr.3: 149572995-149572995 on Build GRCh38
rs368081804 Chr.3: 149572925-149572925 on Build GRCh38

View Full Product Details