Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 110632479-110632722 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TCTN1
Entrez Gene ID
79600
Gene Name
tectonic family member 1
Gene Aliases
JBTS13, TECT1
UniGene
Hs.211511
Gene Chromosome Location
Chr.12: 110614027-110649130 on Build GRCh38
Transcripts
NM_001082537.2 NM_001082538.2 NM_001173975.2 NM_001173976.1 NM_001319680.1 NM_001319681.1 NM_024549.5 NR_135088.1 XM_005253934.3 XM_005253935.3 XM_006719594.2 XM_006719595.2 XM_006719596.2 XM_006719597.3 XM_006719598.2 XM_006719599.2 XM_006719600.2 XM_011538733.2 XM_011538734.2 XM_011538735.1 XM_011538737.2 XM_011538738.2 XM_017019964.1 XM_017019965.1 XM_017019966.1 XM_017019967.1 XM_017019968.1 XM_017019969.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6267035 c.690G>T p.L230L Chr.12: 110632537-110632537 on Build GRCh38
5909308 c.653C>T p.S218L Chr.12: 110632500-110632500 on Build GRCh38
7428789 c.635_636delCT p.L213fs*20 Chr.12: 110632482-110632483 on Build GRCh38
5567703 c.654G>A p.S218S Chr.12: 110632501-110632501 on Build GRCh38
692151 c.709G>C p.A237P Chr.12: 110632556-110632556 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753676064 Chr.12: 110632575-110632575 on Build GRCh38
rs752181429 Chr.12: 110632552-110632552 on Build GRCh38
rs745811491 Chr.12: 110632601-110632601 on Build GRCh38
rs781163162 Chr.12: 110632611-110632611 on Build GRCh38
rs770701543 Chr.12: 110632623-110632623 on Build GRCh38
rs797046038 Chr.12: 110632549-110632551 on Build GRCh38
rs549439598 Chr.12: 110632661-110632661 on Build GRCh38
rs200863258 Chr.12: 110632560-110632560 on Build GRCh38
rs376544711 Chr.12: 110632518-110632518 on Build GRCh38
rs768857454 Chr.12: 110632602-110632602 on Build GRCh38
rs759139558 Chr.12: 110632539-110632539 on Build GRCh38
rs545897929 Chr.12: 110632654-110632654 on Build GRCh38
rs913810931 Chr.12: 110632685-110632685 on Build GRCh38
rs780313300 Chr.12: 110632595-110632595 on Build GRCh38
rs770471504 Chr.12: 110632629-110632629 on Build GRCh38
rs753777109 Chr.12: 110632649-110632649 on Build GRCh38
rs1026171628 Chr.12: 110632610-110632610 on Build GRCh38
rs766946084 Chr.12: 110632548-110632548 on Build GRCh38
rs563888954 Chr.12: 110632678-110632678 on Build GRCh38
rs765317121 Chr.12: 110632662-110632662 on Build GRCh38
rs770297289 Chr.12: 110632514-110632514 on Build GRCh38
rs773823263 Chr.12: 110632630-110632630 on Build GRCh38
rs776715588 Chr.12: 110632501-110632501 on Build GRCh38
rs988502533 Chr.12: 110632681-110632681 on Build GRCh38
rs972727791 Chr.12: 110632603-110632603 on Build GRCh38
rs766174385 Chr.12: 110632521-110632521 on Build GRCh38
rs748208619 Chr.12: 110632617-110632617 on Build GRCh38
rs763386832 Chr.12: 110632519-110632519 on Build GRCh38
rs751252267 Chr.12: 110632530-110632530 on Build GRCh38
rs146815356 Chr.12: 110632613-110632614 on Build GRCh38

View Full Product Details