Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 73864940-73865199 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WBSCR28
Entrez Gene ID
135886
Gene Name
Williams-Beuren syndrome chromosome region 28
Gene Aliases
-
UniGene
Hs.647026
Gene Chromosome Location
Chr.7: 73860848-73865893 on Build GRCh38
Transcripts
NM_182504.3 XM_011515785.2 XM_017011741.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4886256 c.223C>A p.Q75K Chr.7: 73865143-73865143 on Build GRCh38
7168872 c.114C>T p.L38L Chr.7: 73865034-73865034 on Build GRCh38
6821416 c.164delG p.S57fs*27 Chr.7: 73865084-73865084 on Build GRCh38
3685213 c.73-4G>T p.? Chr.7: 73864989-73864989 on Build GRCh38
7227375 c.232T>C p.W78R Chr.7: 73865152-73865152 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782171963 Chr.7: 73865097-73865097 on Build GRCh38
rs782164369 Chr.7: 73865178-73865178 on Build GRCh38
rs782138484 Chr.7: 73865114-73865114 on Build GRCh38
rs781911600 Chr.7: 73864971-73864972 on Build GRCh38
rs782638216 Chr.7: 73865048-73865048 on Build GRCh38
rs781996746 Chr.7: 73865005-73865005 on Build GRCh38
rs530354825 Chr.7: 73865104-73865104 on Build GRCh38
rs150108862 Chr.7: 73865132-73865133 on Build GRCh38
rs782056334 Chr.7: 73865016-73865016 on Build GRCh38
rs558082896 Chr.7: 73865140-73865140 on Build GRCh38
rs375181504 Chr.7: 73865149-73865149 on Build GRCh38
rs782722944 Chr.7: 73865017-73865017 on Build GRCh38
rs17852792 Chr.7: 73865129-73865129 on Build GRCh38
rs373186259 Chr.7: 73865100-73865100 on Build GRCh38
rs782112426 Chr.7: 73865029-73865029 on Build GRCh38
rs145787430 Chr.7: 73865075-73865075 on Build GRCh38
rs574404510 Chr.7: 73864971-73864971 on Build GRCh38
rs782637234 Chr.7: 73865141-73865141 on Build GRCh38
rs528240299 Chr.7: 73865040-73865040 on Build GRCh38
rs782658274 Chr.7: 73865043-73865043 on Build GRCh38
rs782763050 Chr.7: 73865105-73865105 on Build GRCh38
rs782348506 Chr.7: 73865157-73865157 on Build GRCh38
rs782023575 Chr.7: 73865173-73865173 on Build GRCh38
rs782602663 Chr.7: 73864981-73864981 on Build GRCh38
rs782797439 Chr.7: 73865119-73865119 on Build GRCh38
rs782410314 Chr.7: 73865039-73865039 on Build GRCh38
rs202209070 Chr.7: 73865013-73865013 on Build GRCh38
rs782390348 Chr.7: 73865164-73865164 on Build GRCh38
rs781917010 Chr.7: 73865014-73865014 on Build GRCh38
rs782472871 Chr.7: 73865133-73865133 on Build GRCh38
rs374973138 Chr.7: 73864967-73864967 on Build GRCh38
rs782378942 Chr.7: 73864999-73864999 on Build GRCh38
rs56933025 Chr.7: 73865120-73865120 on Build GRCh38
rs782233606 Chr.7: 73864995-73864995 on Build GRCh38
rs781823928 Chr.7: 73865042-73865042 on Build GRCh38
rs781813177 Chr.7: 73865133-73865133 on Build GRCh38
rs201872219 Chr.7: 73865046-73865046 on Build GRCh38
rs782267061 Chr.7: 73865059-73865059 on Build GRCh38
rs375233429 Chr.7: 73865032-73865032 on Build GRCh38
rs200271124 Chr.7: 73865006-73865006 on Build GRCh38
rs367781387 Chr.7: 73864966-73864966 on Build GRCh38
rs201563407 Chr.7: 73865154-73865154 on Build GRCh38
rs61742124 Chr.7: 73865031-73865031 on Build GRCh38
rs782113309 Chr.7: 73865162-73865162 on Build GRCh38
rs569591122 Chr.7: 73865123-73865123 on Build GRCh38
rs202000075 Chr.7: 73865153-73865153 on Build GRCh38
rs781961272 Chr.7: 73865027-73865027 on Build GRCh38
rs199701663 Chr.7: 73864979-73864979 on Build GRCh38
rs377392877 Chr.7: 73865110-73865110 on Build GRCh38
rs782687168 Chr.7: 73865069-73865069 on Build GRCh38
rs782535258 Chr.7: 73865065-73865065 on Build GRCh38
rs13227841 Chr.7: 73865152-73865152 on Build GRCh38
rs781994281 Chr.7: 73865093-73865093 on Build GRCh38
rs782695360 Chr.7: 73865180-73865180 on Build GRCh38
rs374594622 Chr.7: 73865158-73865158 on Build GRCh38

View Full Product Details