Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 6118115-6118342 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WSCD1
Entrez Gene ID
23302
Gene Name
WSC domain containing 1
Gene Aliases
-
UniGene
Hs.370166
Gene Chromosome Location
Chr.17: 6069106-6124427 on Build GRCh38
Transcripts
NM_015253.1 XM_005256572.1 XM_005256573.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1135978 c.1358G>A p.R453H Chr.17: 6118171-6118171 on Build GRCh38
4463727 c.1302C>T p.S434S Chr.17: 6118115-6118115 on Build GRCh38
6081854 c.1328G>C p.C443S Chr.17: 6118141-6118141 on Build GRCh38
706553 c.1309G>T p.A437S Chr.17: 6118122-6118122 on Build GRCh38
296060 c.1375G>T p.E459* Chr.17: 6118188-6118188 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs755222817 Chr.17: 6118232-6118232 on Build GRCh38
rs751601693 Chr.17: 6118154-6118154 on Build GRCh38
rs758576288 Chr.17: 6118146-6118146 on Build GRCh38
rs751894280 Chr.17: 6118228-6118228 on Build GRCh38
rs942902294 Chr.17: 6118216-6118216 on Build GRCh38
rs911291670 Chr.17: 6118191-6118191 on Build GRCh38
rs750750198 Chr.17: 6118141-6118141 on Build GRCh38
rs951057709 Chr.17: 6118320-6118320 on Build GRCh38
rs987306689 Chr.17: 6118173-6118173 on Build GRCh38
rs775152628 Chr.17: 6118200-6118200 on Build GRCh38
rs371850027 Chr.17: 6118205-6118205 on Build GRCh38
rs1043802875 Chr.17: 6118156-6118156 on Build GRCh38
rs368910216 Chr.17: 6118206-6118206 on Build GRCh38
rs771629357 Chr.17: 6118197-6118197 on Build GRCh38
rs765735871 Chr.17: 6118207-6118207 on Build GRCh38
rs879084789 Chr.17: 6118189-6118189 on Build GRCh38
rs773583789 Chr.17: 6118170-6118170 on Build GRCh38
rs186409117 Chr.17: 6118220-6118220 on Build GRCh38
rs781419783 Chr.17: 6118158-6118158 on Build GRCh38
rs1047276143 Chr.17: 6118302-6118302 on Build GRCh38
rs561025043 Chr.17: 6118299-6118299 on Build GRCh38
rs112285134 Chr.17: 6118171-6118171 on Build GRCh38
rs1025732457 Chr.17: 6118307-6118307 on Build GRCh38
rs745637204 Chr.17: 6118184-6118184 on Build GRCh38
rs796648553 Chr.17: 6118255-6118255 on Build GRCh38
rs749281852 Chr.17: 6118161-6118161 on Build GRCh38
rs746653739 Chr.17: 6118202-6118202 on Build GRCh38
rs878932750 Chr.17: 6118285-6118285 on Build GRCh38
rs898791950 Chr.17: 6118234-6118234 on Build GRCh38
rs755165305 Chr.17: 6118155-6118155 on Build GRCh38
rs543882810 Chr.17: 6118254-6118254 on Build GRCh38
rs2302838 Chr.17: 6118221-6118221 on Build GRCh38
rs867251540 Chr.17: 6118144-6118144 on Build GRCh38

View Full Product Details