Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 32543075-32543336 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TOPORS
Entrez Gene ID
10210
Gene Name
TOP1 binding arginine/serine rich protein
Gene Aliases
LUN, P53BP3, RP31, TP53BPL
UniGene
Hs.589962
Gene Chromosome Location
Chr.9: 32540544-32552628 on Build GRCh38
Transcripts
NM_001195622.1 NM_005802.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1314750 c.1282T>G p.S428A Chr.9: 32543243-32543243 on Build GRCh38
5467675 c.1321A>G p.T441A Chr.9: 32543204-32543204 on Build GRCh38
2772112 c.1445G>T p.G482V Chr.9: 32543080-32543080 on Build GRCh38
3656822 c.1348G>A p.V450I Chr.9: 32543177-32543177 on Build GRCh38
4735356 c.1297G>A p.V433I Chr.9: 32543228-32543228 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs544253165 Chr.9: 32543227-32543227 on Build GRCh38
rs61758063 Chr.9: 32543136-32543136 on Build GRCh38
rs958778754 Chr.9: 32543162-32543162 on Build GRCh38
rs770140141 Chr.9: 32543246-32543247 on Build GRCh38
rs905349708 Chr.9: 32543213-32543213 on Build GRCh38
rs575026005 Chr.9: 32543204-32543204 on Build GRCh38
rs962905617 Chr.9: 32543305-32543305 on Build GRCh38
rs368302118 Chr.9: 32543218-32543218 on Build GRCh38
rs540717992 Chr.9: 32543234-32543234 on Build GRCh38
rs780866628 Chr.9: 32543160-32543160 on Build GRCh38
rs774925467 Chr.9: 32543166-32543166 on Build GRCh38
rs61758062 Chr.9: 32543287-32543287 on Build GRCh38
rs201027247 Chr.9: 32543198-32543198 on Build GRCh38
rs780802720 Chr.9: 32543289-32543289 on Build GRCh38
rs764067914 Chr.9: 32543265-32543265 on Build GRCh38
rs779750604 Chr.9: 32543303-32543303 on Build GRCh38
rs777634455 Chr.9: 32543137-32543137 on Build GRCh38
rs745677183 Chr.9: 32543290-32543290 on Build GRCh38
rs755081657 Chr.9: 32543129-32543129 on Build GRCh38
rs758608005 Chr.9: 32543256-32543256 on Build GRCh38
rs769459711 Chr.9: 32543164-32543164 on Build GRCh38
rs765558710 Chr.9: 32543243-32543243 on Build GRCh38
rs746241882 Chr.9: 32543116-32543118 on Build GRCh38
rs755699342 Chr.9: 32543295-32543295 on Build GRCh38
rs372508949 Chr.9: 32543310-32543310 on Build GRCh38
rs751491112 Chr.9: 32543280-32543280 on Build GRCh38
rs768103328 Chr.9: 32543311-32543311 on Build GRCh38
rs752817816 Chr.9: 32543248-32543248 on Build GRCh38
rs770678902 Chr.9: 32543152-32543152 on Build GRCh38
rs148416446 Chr.9: 32543134-32543134 on Build GRCh38
rs910122958 Chr.9: 32543304-32543304 on Build GRCh38
rs772566291 Chr.9: 32543200-32543200 on Build GRCh38
rs745481592 Chr.9: 32543161-32543161 on Build GRCh38
rs200877916 Chr.9: 32543228-32543228 on Build GRCh38
rs146483990 Chr.9: 32543146-32543146 on Build GRCh38
rs746804680 Chr.9: 32543139-32543139 on Build GRCh38
rs1013645856 Chr.9: 32543144-32543144 on Build GRCh38
rs769934695 Chr.9: 32543222-32543222 on Build GRCh38

View Full Product Details