Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 110387138-110387393 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SVEP1
Entrez Gene ID
79987
Gene Name
sushi, von Willebrand factor type A, EGF and pentraxin domain containing 1
Gene Aliases
C9orf13, CCP22, POLYDOM, SEL-OB, SELOB
UniGene
Hs.522334
Gene Chromosome Location
Chr.9: 110365248-110579880 on Build GRCh38
Transcripts
NM_153366.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3652811 c.10048C>T p.P3350S Chr.9: 110387297-110387297 on Build GRCh38
3779535 c.9988A>G p.S3330G Chr.9: 110387357-110387357 on Build GRCh38
3169398 c.9956C>A p.P3319H Chr.9: 110387389-110387389 on Build GRCh38
86710 c.10037G>A p.S3346N Chr.9: 110387308-110387308 on Build GRCh38
4816655 c.9971C>T p.S3324F Chr.9: 110387374-110387374 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs909150481 Chr.9: 110387214-110387214 on Build GRCh38
rs771382502 Chr.9: 110387333-110387333 on Build GRCh38
rs372409135 Chr.9: 110387357-110387357 on Build GRCh38
rs774690333 Chr.9: 110387335-110387335 on Build GRCh38
rs980573618 Chr.9: 110387212-110387212 on Build GRCh38
rs200311711 Chr.9: 110387281-110387281 on Build GRCh38
rs142463374 Chr.9: 110387328-110387328 on Build GRCh38
rs759190053 Chr.9: 110387340-110387340 on Build GRCh38
rs768933750 Chr.9: 110387296-110387296 on Build GRCh38
rs753665110 Chr.9: 110387347-110387347 on Build GRCh38
rs746170979 Chr.9: 110387334-110387334 on Build GRCh38
rs921472934 Chr.9: 110387227-110387227 on Build GRCh38
rs767077311 Chr.9: 110387256-110387256 on Build GRCh38
rs977536075 Chr.9: 110387195-110387195 on Build GRCh38
rs967078127 Chr.9: 110387186-110387186 on Build GRCh38
rs777716373 Chr.9: 110387269-110387269 on Build GRCh38
rs756018074 Chr.9: 110387268-110387268 on Build GRCh38
rs763898532 Chr.9: 110387285-110387285 on Build GRCh38
rs781439987 Chr.9: 110387317-110387317 on Build GRCh38
rs368624561 Chr.9: 110387239-110387239 on Build GRCh38
rs559268210 Chr.9: 110387240-110387240 on Build GRCh38
rs568255918 Chr.9: 110387187-110387187 on Build GRCh38
rs553797332 Chr.9: 110387203-110387203 on Build GRCh38
rs778561103 Chr.9: 110387284-110387284 on Build GRCh38
rs544465911 Chr.9: 110387324-110387324 on Build GRCh38
rs969216307 Chr.9: 110387201-110387201 on Build GRCh38
rs1051462695 Chr.9: 110387263-110387263 on Build GRCh38
rs753872713 Chr.9: 110387358-110387358 on Build GRCh38
rs114581956 Chr.9: 110387332-110387332 on Build GRCh38
rs770237687 Chr.9: 110387325-110387325 on Build GRCh38
rs752136641 Chr.9: 110387265-110387265 on Build GRCh38
rs577484502 Chr.9: 110387273-110387273 on Build GRCh38
rs943557513 Chr.9: 110387275-110387275 on Build GRCh38
rs201533942 Chr.9: 110387346-110387346 on Build GRCh38
rs137911727 Chr.9: 110387215-110387215 on Build GRCh38
rs767097945 Chr.9: 110387343-110387343 on Build GRCh38
rs933105750 Chr.9: 110387250-110387250 on Build GRCh38
rs79932775 Chr.9: 110387228-110387228 on Build GRCh38
rs371736321 Chr.9: 110387163-110387163 on Build GRCh38
rs759171842 Chr.9: 110387254-110387254 on Build GRCh38

View Full Product Details