Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41708649-41708904 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEACAM5
Entrez Gene ID
1048
Gene Name
carcinoembryonic antigen related cell adhesion molecule 5
Gene Aliases
CD66e, CEA
UniGene
Hs.709196
Gene Chromosome Location
Chr.19: 41708585-41730433 on Build GRCh38
Transcripts
NM_001291484.2 NM_001308398.1 NM_004363.5 XM_005258413.3 XM_011526322.1 XM_017026145.1 XM_017026146.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5389877 c.11C>T p.P4L Chr.19: 41708742-41708742 on Build GRCh38
4911888 c.3G>A p.M1I Chr.19: 41708734-41708734 on Build GRCh38
5669316 c.14C>T p.S5L Chr.19: 41708745-41708745 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs201518445 Chr.19: 41708745-41708745 on Build GRCh38
rs781952967 Chr.19: 41708735-41708735 on Build GRCh38
rs782408845 Chr.19: 41708828-41708828 on Build GRCh38
rs781992451 Chr.19: 41708706-41708706 on Build GRCh38
rs888173494 Chr.19: 41708785-41708785 on Build GRCh38
rs782037600 Chr.19: 41708831-41708831 on Build GRCh38
rs538013635 Chr.19: 41708820-41708820 on Build GRCh38
rs782317994 Chr.19: 41708731-41708731 on Build GRCh38
rs782815290 Chr.19: 41708757-41708757 on Build GRCh38
rs782583505 Chr.19: 41708696-41708696 on Build GRCh38
rs782150992 Chr.19: 41708748-41708748 on Build GRCh38
rs781958989 Chr.19: 41708840-41708840 on Build GRCh38
rs782399172 Chr.19: 41708704-41708704 on Build GRCh38
rs782171457 Chr.19: 41708730-41708730 on Build GRCh38
rs782589950 Chr.19: 41708812-41708812 on Build GRCh38
rs267605502 Chr.19: 41708742-41708742 on Build GRCh38
rs781846226 Chr.19: 41708781-41708781 on Build GRCh38
rs781810856 Chr.19: 41708698-41708698 on Build GRCh38
rs781900989 Chr.19: 41708797-41708797 on Build GRCh38
rs200207181 Chr.19: 41708826-41708826 on Build GRCh38
rs1040941041 Chr.19: 41708756-41708756 on Build GRCh38
rs782727394 Chr.19: 41708771-41708771 on Build GRCh38
rs115233397 Chr.19: 41708725-41708725 on Build GRCh38
rs782163428 Chr.19: 41708752-41708753 on Build GRCh38
rs1026803117 Chr.19: 41708777-41708777 on Build GRCh38
rs971363260 Chr.19: 41708675-41708675 on Build GRCh38
rs111973464 Chr.19: 41708834-41708835 on Build GRCh38
rs782333750 Chr.19: 41708832-41708832 on Build GRCh38
rs45614132 Chr.19: 41708703-41708703 on Build GRCh38
rs190377739 Chr.19: 41708702-41708702 on Build GRCh38
rs782429666 Chr.19: 41708811-41708811 on Build GRCh38
rs1042765987 Chr.19: 41708714-41708714 on Build GRCh38
rs782690285 Chr.19: 41708833-41708834 on Build GRCh38
rs200859918 Chr.19: 41708782-41708782 on Build GRCh38
rs45537140 Chr.19: 41708694-41708694 on Build GRCh38
rs144340352 Chr.19: 41708825-41708826 on Build GRCh38
rs114263190 Chr.19: 41708878-41708878 on Build GRCh38
rs72455487 Chr.19: 41708835-41708836 on Build GRCh38
rs1015646491 Chr.19: 41708858-41708858 on Build GRCh38
rs782487516 Chr.19: 41708701-41708701 on Build GRCh38
rs781944250 Chr.19: 41708843-41708843 on Build GRCh38
rs782101286 Chr.19: 41708841-41708841 on Build GRCh38
rs782342901 Chr.19: 41708869-41708869 on Build GRCh38
rs3217253 Chr.19: 41708836-41708837 on Build GRCh38
rs200391100 Chr.19: 41708770-41708770 on Build GRCh38

View Full Product Details