Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 55077176-55077401 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
USP24
Entrez Gene ID
23358
Gene Name
ubiquitin specific peptidase 24
Gene Aliases
-
UniGene
Hs.477009
Gene Chromosome Location
Chr.1: 55066359-55215374 on Build GRCh38
Transcripts
NM_015306.2 XM_005270690.3 XM_017000831.1 XM_017000832.1 XM_017000833.1 XM_017000834.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7410736 c.7354A>G p.T2452A Chr.1: 55077261-55077261 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs781352610 Chr.1: 55077278-55077278 on Build GRCh38
rs1040554962 Chr.1: 55077345-55077345 on Build GRCh38
rs769193274 Chr.1: 55077307-55077307 on Build GRCh38
rs756647895 Chr.1: 55077287-55077287 on Build GRCh38
rs373685319 Chr.1: 55077343-55077343 on Build GRCh38
rs12568453 Chr.1: 55077234-55077234 on Build GRCh38
rs1021669279 Chr.1: 55077327-55077327 on Build GRCh38
rs17111576 Chr.1: 55077295-55077295 on Build GRCh38
rs763850440 Chr.1: 55077245-55077245 on Build GRCh38
rs757428170 Chr.1: 55077259-55077259 on Build GRCh38
rs745405079 Chr.1: 55077371-55077371 on Build GRCh38
rs185353600 Chr.1: 55077286-55077286 on Build GRCh38
rs866884740 Chr.1: 55077260-55077260 on Build GRCh38
rs752374149 Chr.1: 55077227-55077227 on Build GRCh38
rs751815641 Chr.1: 55077253-55077253 on Build GRCh38
rs780492160 Chr.1: 55077294-55077294 on Build GRCh38
rs1011663380 Chr.1: 55077320-55077320 on Build GRCh38
rs779391881 Chr.1: 55077328-55077328 on Build GRCh38
rs930862963 Chr.1: 55077221-55077221 on Build GRCh38

View Full Product Details