Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 75813510-75813783 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCL24
Entrez Gene ID
6369
Gene Name
C-C motif chemokine ligand 24
Gene Aliases
Ckb-6, MPIF-2, MPIF2, SCYA24
UniGene
Hs.247838
Gene Chromosome Location
Chr.7: 75810825-75823356 on Build GRCh38
Transcripts
NM_002991.2 XM_011516459.2 XM_011516460.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3412302 c.71C>T p.T24M Chr.7: 75813645-75813645 on Build GRCh38
1698767 c.58C>T p.H20Y Chr.7: 75813658-75813658 on Build GRCh38
6622863 c.50G>A p.C17Y Chr.7: 75813666-75813666 on Build GRCh38
6032282 c.71C>G p.T24R Chr.7: 75813645-75813645 on Build GRCh38
4395842 c.67C>T p.P23S Chr.7: 75813649-75813649 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782611011 Chr.7: 75813742-75813742 on Build GRCh38
rs200638877 Chr.7: 75813610-75813610 on Build GRCh38
rs782141375 Chr.7: 75813697-75813697 on Build GRCh38
rs994805478 Chr.7: 75813579-75813579 on Build GRCh38
rs193245535 Chr.7: 75813722-75813722 on Build GRCh38
rs781862888 Chr.7: 75813721-75813721 on Build GRCh38
rs371344336 Chr.7: 75813745-75813745 on Build GRCh38
rs782028480 Chr.7: 75813690-75813690 on Build GRCh38
rs376218311 Chr.7: 75813759-75813759 on Build GRCh38
rs782807798 Chr.7: 75813621-75813621 on Build GRCh38
rs782315245 Chr.7: 75813707-75813707 on Build GRCh38
rs200262991 Chr.7: 75813589-75813589 on Build GRCh38
rs782054898 Chr.7: 75813623-75813623 on Build GRCh38
rs782441575 Chr.7: 75813616-75813616 on Build GRCh38
rs782085727 Chr.7: 75813712-75813712 on Build GRCh38
rs558400267 Chr.7: 75813571-75813571 on Build GRCh38
rs781810208 Chr.7: 75813622-75813622 on Build GRCh38
rs781818235 Chr.7: 75813744-75813744 on Build GRCh38
rs782434669 Chr.7: 75813741-75813741 on Build GRCh38
rs781810847 Chr.7: 75813732-75813732 on Build GRCh38
rs11465288 Chr.7: 75813632-75813632 on Build GRCh38
rs900792102 Chr.7: 75813546-75813546 on Build GRCh38
rs200038952 Chr.7: 75813645-75813645 on Build GRCh38
rs782812042 Chr.7: 75813726-75813726 on Build GRCh38
rs782749237 Chr.7: 75813625-75813625 on Build GRCh38
rs989134700 Chr.7: 75813672-75813672 on Build GRCh38
rs782594363 Chr.7: 75813680-75813680 on Build GRCh38
rs2302004 Chr.7: 75813537-75813537 on Build GRCh38
rs372946559 Chr.7: 75813636-75813636 on Build GRCh38
rs537428760 Chr.7: 75813550-75813550 on Build GRCh38
rs782257140 Chr.7: 75813686-75813686 on Build GRCh38
rs782781825 Chr.7: 75813716-75813716 on Build GRCh38
rs781822160 Chr.7: 75813629-75813629 on Build GRCh38
rs369778604 Chr.7: 75813593-75813593 on Build GRCh38
rs529320164 Chr.7: 75813708-75813708 on Build GRCh38
rs188963157 Chr.7: 75813644-75813644 on Build GRCh38
rs11465289 Chr.7: 75813615-75813615 on Build GRCh38
rs375638264 Chr.7: 75813631-75813631 on Build GRCh38
rs562220612 Chr.7: 75813687-75813687 on Build GRCh38
rs78198843 Chr.7: 75813620-75813620 on Build GRCh38
rs890252092 Chr.7: 75813752-75813752 on Build GRCh38
rs782355653 Chr.7: 75813648-75813648 on Build GRCh38
rs782625978 Chr.7: 75813758-75813758 on Build GRCh38

View Full Product Details