Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 27773369-27773596 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STX12
Entrez Gene ID
23673
Gene Name
syntaxin 12
Gene Aliases
STX13, STX14
UniGene
Hs.523855
Gene Chromosome Location
Chr.1: 27773183-27824452 on Build GRCh38
Transcripts
NM_177424.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4236370 c.90C>T p.S30S Chr.1: 27773397-27773397 on Build GRCh38
4236371 c.90C>T p.S30S Chr.1: 27773397-27773397 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1010431855 Chr.1: 27773565-27773565 on Build GRCh38
rs1030620918 Chr.1: 27773548-27773548 on Build GRCh38
rs571210293 Chr.1: 27773563-27773563 on Build GRCh38
rs376046525 Chr.1: 27773498-27773498 on Build GRCh38
rs780605440 Chr.1: 27773425-27773425 on Build GRCh38
rs997732583 Chr.1: 27773527-27773527 on Build GRCh38
rs1015963315 Chr.1: 27773550-27773550 on Build GRCh38
rs749960736 Chr.1: 27773410-27773410 on Build GRCh38
rs760617815 Chr.1: 27773394-27773394 on Build GRCh38
rs1038671528 Chr.1: 27773521-27773521 on Build GRCh38
rs758815800 Chr.1: 27773419-27773419 on Build GRCh38
rs756958960 Chr.1: 27773404-27773404 on Build GRCh38
rs747594726 Chr.1: 27773441-27773441 on Build GRCh38
rs548101240 Chr.1: 27773524-27773524 on Build GRCh38
rs929182258 Chr.1: 27773478-27773478 on Build GRCh38
rs375964792 Chr.1: 27773398-27773398 on Build GRCh38
rs755381064 Chr.1: 27773437-27773437 on Build GRCh38
rs771609825 Chr.1: 27773464-27773464 on Build GRCh38
rs369960212 Chr.1: 27773460-27773460 on Build GRCh38
rs769409590 Chr.1: 27773417-27773418 on Build GRCh38
rs113246948 Chr.1: 27773494-27773494 on Build GRCh38
rs777658909 Chr.1: 27773471-27773471 on Build GRCh38
rs753594224 Chr.1: 27773402-27773402 on Build GRCh38
rs868439662 Chr.1: 27773447-27773447 on Build GRCh38
rs887566173 Chr.1: 27773507-27773507 on Build GRCh38
rs1005453552 Chr.1: 27773517-27773517 on Build GRCh38
rs749446736 Chr.1: 27773473-27773473 on Build GRCh38
rs531339904 Chr.1: 27773435-27773435 on Build GRCh38
rs996656579 Chr.1: 27773495-27773504 on Build GRCh38
rs564611237 Chr.1: 27773513-27773513 on Build GRCh38
rs781049024 Chr.1: 27773475-27773475 on Build GRCh38
rs764613166 Chr.1: 27773405-27773405 on Build GRCh38

View Full Product Details