Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 133583855-133584102 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDV3
Entrez Gene ID
55573
Gene Name
CDV3 homolog
Gene Aliases
H41
UniGene
Hs.518265
Gene Chromosome Location
Chr.3: 133573563-133590274 on Build GRCh38
Transcripts
NM_001134422.1 NM_001134423.2 NM_001282762.1 NM_001282763.1 NM_001282764.1 NM_001282765.1 NM_017548.4 XM_005247588.4 XM_005247589.4 XM_005247590.4 XM_005247591.4 XM_005247592.4 XM_005247594.4 XM_011512961.2 XM_011512962.2 XM_017006808.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3122228 c.343G>A p.E115K Chr.3: 133584027-133584027 on Build GRCh38
6407104 c.362G>T p.G121V Chr.3: 133584046-133584046 on Build GRCh38
6401192 c.327G>T p.K109N Chr.3: 133584011-133584011 on Build GRCh38
3122230 c.418C>T p.P140S Chr.3: 133584102-133584102 on Build GRCh38
6362277 c.330A>T p.E110D Chr.3: 133584014-133584014 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs35495859 Chr.3: 133584063-133584063 on Build GRCh38
rs554799281 Chr.3: 133583933-133583933 on Build GRCh38
rs1009117654 Chr.3: 133583921-133583921 on Build GRCh38
rs201293169 Chr.3: 133584072-133584072 on Build GRCh38
rs747077477 Chr.3: 133584036-133584036 on Build GRCh38
rs763768656 Chr.3: 133584017-133584017 on Build GRCh38
rs781566777 Chr.3: 133584047-133584047 on Build GRCh38
rs549915446 Chr.3: 133583891-133583891 on Build GRCh38
rs929960576 Chr.3: 133584003-133584003 on Build GRCh38
rs202089095 Chr.3: 133584056-133584056 on Build GRCh38
rs762296467 Chr.3: 133583952-133583952 on Build GRCh38
rs529967304 Chr.3: 133583890-133583890 on Build GRCh38
rs923682790 Chr.3: 133583908-133583908 on Build GRCh38
rs774916239 Chr.3: 133583977-133583977 on Build GRCh38
rs140108131 Chr.3: 133584020-133584020 on Build GRCh38
rs113870525 Chr.3: 133583999-133583999 on Build GRCh38
rs964764000 Chr.3: 133583930-133583930 on Build GRCh38
rs748463641 Chr.3: 133584064-133584064 on Build GRCh38
rs755208361 Chr.3: 133584037-133584037 on Build GRCh38
rs543844840 Chr.3: 133583953-133583954 on Build GRCh38
rs570072492 Chr.3: 133583914-133583914 on Build GRCh38
rs770355054 Chr.3: 133583960-133583960 on Build GRCh38
rs765072468 Chr.3: 133584029-133584029 on Build GRCh38
rs906484115 Chr.3: 133583985-133583985 on Build GRCh38
rs758306813 Chr.3: 133584032-133584032 on Build GRCh38
rs763439625 Chr.3: 133583975-133583975 on Build GRCh38
rs916436449 Chr.3: 133583942-133583942 on Build GRCh38
rs1054252928 Chr.3: 133583880-133583880 on Build GRCh38
rs767025438 Chr.3: 133583976-133583976 on Build GRCh38
rs199906391 Chr.3: 133584021-133584021 on Build GRCh38
rs181969008 Chr.3: 133583889-133583889 on Build GRCh38
rs1009657125 Chr.3: 133583894-133583894 on Build GRCh38
rs780159708 Chr.3: 133584033-133584033 on Build GRCh38
rs776799673 Chr.3: 133584030-133584030 on Build GRCh38
rs1018008529 Chr.3: 133583926-133583926 on Build GRCh38
rs760327005 Chr.3: 133583987-133583987 on Build GRCh38
rs759103785 Chr.3: 133583915-133583915 on Build GRCh38
rs1021081616 Chr.3: 133583898-133583898 on Build GRCh38
rs753824799 Chr.3: 133584014-133584014 on Build GRCh38
rs773779474 Chr.3: 133583962-133583962 on Build GRCh38

View Full Product Details