Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 64522143-64522370 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF680
Entrez Gene ID
340252
Gene Name
zinc finger protein 680
Gene Aliases
-
UniGene
Hs.520886
Gene Chromosome Location
Chr.7: 64500253-64563135 on Build GRCh38
Transcripts
NM_178558.4 XM_006715959.2 XM_017012126.1 XM_017012127.1 XM_017012128.1 XM_017012129.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6033735 c.598C>T p.H200Y Chr.7: 64522156-64522156 on Build GRCh38
6262572 c.510C>A p.N170K Chr.7: 64522244-64522244 on Build GRCh38
6416968 c.533G>A p.G178E Chr.7: 64522221-64522221 on Build GRCh38
6264206 c.477C>T p.Y159Y Chr.7: 64522277-64522277 on Build GRCh38
6263734 c.513T>A p.S171R Chr.7: 64522241-64522241 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775294496 Chr.7: 64522303-64522303 on Build GRCh38
rs748818726 Chr.7: 64522245-64522250 on Build GRCh38
rs764310845 Chr.7: 64522224-64522224 on Build GRCh38
rs771911311 Chr.7: 64522302-64522302 on Build GRCh38
rs375747442 Chr.7: 64522195-64522195 on Build GRCh38
rs1024166128 Chr.7: 64522210-64522210 on Build GRCh38
rs765358421 Chr.7: 64522322-64522322 on Build GRCh38
rs1016269763 Chr.7: 64522187-64522187 on Build GRCh38
rs760888011 Chr.7: 64522222-64522222 on Build GRCh38
rs750986757 Chr.7: 64522242-64522242 on Build GRCh38
rs571429645 Chr.7: 64522277-64522277 on Build GRCh38
rs531055436 Chr.7: 64522276-64522276 on Build GRCh38
rs972707936 Chr.7: 64522251-64522251 on Build GRCh38
rs759234510 Chr.7: 64522192-64522192 on Build GRCh38
rs760305944 Chr.7: 64522287-64522287 on Build GRCh38
rs772792138 Chr.7: 64522178-64522178 on Build GRCh38
rs757354644 Chr.7: 64522236-64522236 on Build GRCh38
rs769775041 Chr.7: 64522228-64522230 on Build GRCh38
rs752861421 Chr.7: 64522206-64522206 on Build GRCh38
rs758872230 Chr.7: 64522329-64522329 on Build GRCh38
rs776874015 Chr.7: 64522320-64522320 on Build GRCh38
rs766819322 Chr.7: 64522333-64522333 on Build GRCh38
rs77820012 Chr.7: 64522248-64522248 on Build GRCh38
rs373687231 Chr.7: 64522274-64522274 on Build GRCh38
rs200167008 Chr.7: 64522190-64522190 on Build GRCh38
rs867520358 Chr.7: 64522201-64522201 on Build GRCh38
rs780531809 Chr.7: 64522246-64522246 on Build GRCh38
rs146417840 Chr.7: 64522245-64522245 on Build GRCh38
rs775486364 Chr.7: 64522243-64522246 on Build GRCh38
rs376585361 Chr.7: 64522296-64522296 on Build GRCh38
rs765702238 Chr.7: 64522239-64522239 on Build GRCh38
rs760482124 Chr.7: 64522304-64522304 on Build GRCh38
rs751914040 Chr.7: 64522338-64522338 on Build GRCh38
rs199754812 Chr.7: 64522275-64522275 on Build GRCh38
rs958668020 Chr.7: 64522180-64522180 on Build GRCh38
rs762576067 Chr.7: 64522179-64522179 on Build GRCh38
rs753862039 Chr.7: 64522227-64522227 on Build GRCh38
rs370045379 Chr.7: 64522261-64522261 on Build GRCh38
rs761859515 Chr.7: 64522321-64522321 on Build GRCh38
rs770953671 Chr.7: 64522181-64522181 on Build GRCh38
rs775905560 Chr.7: 64522259-64522259 on Build GRCh38
rs768145929 Chr.7: 64522320-64522320 on Build GRCh38
rs144009359 Chr.7: 64522177-64522177 on Build GRCh38
rs750887660 Chr.7: 64522323-64522323 on Build GRCh38
rs140933799 Chr.7: 64522255-64522255 on Build GRCh38
rs755433540 Chr.7: 64522341-64522341 on Build GRCh38
rs201430583 Chr.7: 64522315-64522315 on Build GRCh38

View Full Product Details