Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127716731-127716968 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTC16
Entrez Gene ID
158248
Gene Name
tetratricopeptide repeat domain 16
Gene Aliases
-
UniGene
Hs.642748
Gene Chromosome Location
Chr.9: 127716064-127731600 on Build GRCh38
Transcripts
NM_001317037.1 NM_144965.2 XM_005251733.1 XM_005251734.1 XM_006716970.1 XM_006716971.1 XM_011518283.1 XM_011518284.1 XM_011518285.1 XM_011518288.1 XM_011518289.2 XM_011518290.1 XM_011518291.1 XM_011518292.1 XM_011518293.1 XM_011518294.2 XM_011518295.1 XM_011518297.1 XM_011518298.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7113107 c.86A>G p.K29R Chr.9: 127716911-127716911 on Build GRCh38
6115173 c.104T>A p.I35N Chr.9: 127716929-127716929 on Build GRCh38
5518913 c.121G>A p.V41M Chr.9: 127716946-127716946 on Build GRCh38
4804251 c.120C>A p.H40Q Chr.9: 127716945-127716945 on Build GRCh38
328257 c.76C>G p.P26A Chr.9: 127716901-127716901 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139574252 Chr.9: 127716926-127716926 on Build GRCh38
rs1056757310 Chr.9: 127716764-127716764 on Build GRCh38
rs762925689 Chr.9: 127716883-127716883 on Build GRCh38
rs755455482 Chr.9: 127716936-127716936 on Build GRCh38
rs549212049 Chr.9: 127716799-127716799 on Build GRCh38
rs879173693 Chr.9: 127716796-127716796 on Build GRCh38
rs781634912 Chr.9: 127716803-127716803 on Build GRCh38
rs766793641 Chr.9: 127716934-127716934 on Build GRCh38
rs776374543 Chr.9: 127716846-127716846 on Build GRCh38
rs777281075 Chr.9: 127716817-127716817 on Build GRCh38
rs746608995 Chr.9: 127716832-127716832 on Build GRCh38
rs903899672 Chr.9: 127716867-127716867 on Build GRCh38
rs769583810 Chr.9: 127716862-127716862 on Build GRCh38
rs555970103 Chr.9: 127716777-127716777 on Build GRCh38
rs1032188374 Chr.9: 127716933-127716933 on Build GRCh38
rs747582342 Chr.9: 127716810-127716810 on Build GRCh38
rs756587620 Chr.9: 127716932-127716932 on Build GRCh38
rs759372121 Chr.9: 127716856-127716856 on Build GRCh38
rs754429655 Chr.9: 127716935-127716935 on Build GRCh38
rs562917588 Chr.9: 127716909-127716909 on Build GRCh38
rs916327047 Chr.9: 127716774-127716774 on Build GRCh38
rs73668586 Chr.9: 127716911-127716911 on Build GRCh38
rs771681074 Chr.9: 127716815-127716815 on Build GRCh38
rs777274157 Chr.9: 127716946-127716946 on Build GRCh38
rs114770292 Chr.9: 127716931-127716931 on Build GRCh38
rs762086277 Chr.9: 127716914-127716914 on Build GRCh38
rs756640860 Chr.9: 127716807-127716807 on Build GRCh38
rs761369792 Chr.9: 127716875-127716875 on Build GRCh38
rs368982275 Chr.9: 127716836-127716836 on Build GRCh38
rs757564332 Chr.9: 127716802-127716802 on Build GRCh38
rs371264748 Chr.9: 127716808-127716808 on Build GRCh38
rs748136258 Chr.9: 127716946-127716946 on Build GRCh38

View Full Product Details