Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 15975175-15975400 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADORA2B
Entrez Gene ID
136
Gene Name
adenosine A2b receptor
Gene Aliases
ADORA2
UniGene
Hs.167046
Gene Chromosome Location
Chr.17: 15927781-15975896 on Build GRCh38
Transcripts
NM_000676.2 XM_011523659.2 XM_017024197.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5487494 c.877C>T p.R293W Chr.17: 15975220-15975220 on Build GRCh38
3514546 c.878G>A p.R293Q Chr.17: 15975221-15975221 on Build GRCh38
6595325 c.959G>A p.G320D Chr.17: 15975302-15975302 on Build GRCh38
3514547 c.982C>T p.L328F Chr.17: 15975325-15975325 on Build GRCh38
1381091 c.840T>C p.H280H Chr.17: 15975183-15975183 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757998635 Chr.17: 15975229-15975229 on Build GRCh38
rs372636597 Chr.17: 15975291-15975291 on Build GRCh38
rs756524360 Chr.17: 15975322-15975322 on Build GRCh38
rs764654008 Chr.17: 15975302-15975302 on Build GRCh38
rs150639935 Chr.17: 15975281-15975281 on Build GRCh38
rs377355316 Chr.17: 15975353-15975353 on Build GRCh38
rs754401984 Chr.17: 15975327-15975327 on Build GRCh38
rs780001347 Chr.17: 15975372-15975373 on Build GRCh38
rs1021224191 Chr.17: 15975221-15975221 on Build GRCh38
rs1036876439 Chr.17: 15975358-15975358 on Build GRCh38
rs540571442 Chr.17: 15975240-15975240 on Build GRCh38
rs757312723 Chr.17: 15975255-15975255 on Build GRCh38
rs555538647 Chr.17: 15975219-15975219 on Build GRCh38
rs757726498 Chr.17: 15975328-15975328 on Build GRCh38
rs753197314 Chr.17: 15975363-15975365 on Build GRCh38
rs138464210 Chr.17: 15975220-15975220 on Build GRCh38
rs772327012 Chr.17: 15975263-15975263 on Build GRCh38
rs750229596 Chr.17: 15975355-15975355 on Build GRCh38
rs764503783 Chr.17: 15975323-15975323 on Build GRCh38
rs898452175 Chr.17: 15975367-15975367 on Build GRCh38
rs768633790 Chr.17: 15975277-15975277 on Build GRCh38
rs371657178 Chr.17: 15975236-15975236 on Build GRCh38
rs200741295 Chr.17: 15975230-15975230 on Build GRCh38
rs753096102 Chr.17: 15975235-15975235 on Build GRCh38
rs746092412 Chr.17: 15975262-15975262 on Build GRCh38
rs761703634 Chr.17: 15975294-15975295 on Build GRCh38
rs762569718 Chr.17: 15975306-15975306 on Build GRCh38
rs762496834 Chr.17: 15975271-15975271 on Build GRCh38
rs544202480 Chr.17: 15975227-15975227 on Build GRCh38
rs199872870 Chr.17: 15975317-15975317 on Build GRCh38
rs77664279 Chr.17: 15975243-15975243 on Build GRCh38
rs758586452 Chr.17: 15975372-15975372 on Build GRCh38
rs764017942 Chr.17: 15975211-15975211 on Build GRCh38
rs754032994 Chr.17: 15975282-15975283 on Build GRCh38
rs55707398 Chr.17: 15975290-15975290 on Build GRCh38
rs778919081 Chr.17: 15975339-15975339 on Build GRCh38
rs778984983 Chr.17: 15975257-15975257 on Build GRCh38
rs141912284 Chr.17: 15975266-15975266 on Build GRCh38
rs766049607 Chr.17: 15975310-15975310 on Build GRCh38
rs765190357 Chr.17: 15975307-15975308 on Build GRCh38
rs878874119 Chr.17: 15975204-15975204 on Build GRCh38
rs760665957 Chr.17: 15975206-15975206 on Build GRCh38
rs772717092 Chr.17: 15975305-15975305 on Build GRCh38

View Full Product Details