Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 3810657-3810930 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZFR2
Entrez Gene ID
23217
Gene Name
zinc finger RNA binding protein 2
Gene Aliases
KIAA1086
UniGene
Hs.65750
Gene Chromosome Location
Chr.19: 3804024-3869029 on Build GRCh38
Transcripts
NM_015174.1 XM_011527831.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6043185 c.2425C>T p.P809S Chr.19: 3810758-3810758 on Build GRCh38
7338023 c.2366T>G p.V789G Chr.19: 3810817-3810817 on Build GRCh38
6403493 c.2407C>G p.P803A Chr.19: 3810776-3810776 on Build GRCh38
6245558 c.2397C>A p.C799* Chr.19: 3810786-3810786 on Build GRCh38
4638732 c.2413T>A p.W805R Chr.19: 3810770-3810770 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375058331 Chr.19: 3810841-3810841 on Build GRCh38
rs367644907 Chr.19: 3810785-3810785 on Build GRCh38
rs566696286 Chr.19: 3810767-3810767 on Build GRCh38
rs568297927 Chr.19: 3810744-3810744 on Build GRCh38
rs1010401913 Chr.19: 3810911-3810911 on Build GRCh38
rs765495665 Chr.19: 3810865-3810865 on Build GRCh38
rs754128578 Chr.19: 3810850-3810850 on Build GRCh38
rs552322198 Chr.19: 3810781-3810781 on Build GRCh38
rs538094451 Chr.19: 3810834-3810834 on Build GRCh38
rs767595355 Chr.19: 3810833-3810833 on Build GRCh38
rs574879784 Chr.19: 3810851-3810851 on Build GRCh38
rs770388068 Chr.19: 3810842-3810842 on Build GRCh38
rs749512207 Chr.19: 3810736-3810736 on Build GRCh38
rs778953395 Chr.19: 3810866-3810866 on Build GRCh38
rs367762654 Chr.19: 3810850-3810850 on Build GRCh38
rs751131550 Chr.19: 3810818-3810818 on Build GRCh38
rs565330597 Chr.19: 3810764-3810764 on Build GRCh38
rs12610211 Chr.19: 3810711-3810711 on Build GRCh38
rs996805187 Chr.19: 3810896-3810896 on Build GRCh38
rs577231395 Chr.19: 3810904-3810904 on Build GRCh38
rs772104982 Chr.19: 3810895-3810895 on Build GRCh38
rs769259870 Chr.19: 3810782-3810782 on Build GRCh38
rs766576842 Chr.19: 3810805-3810806 on Build GRCh38
rs76514484 Chr.19: 3810727-3810727 on Build GRCh38
rs774490788 Chr.19: 3810740-3810740 on Build GRCh38
rs762666960 Chr.19: 3810784-3810784 on Build GRCh38
rs190072850 Chr.19: 3810804-3810804 on Build GRCh38
rs534329674 Chr.19: 3810886-3810886 on Build GRCh38
rs138287525 Chr.19: 3810775-3810775 on Build GRCh38
rs937215541 Chr.19: 3810819-3810819 on Build GRCh38
rs955155457 Chr.19: 3810906-3810906 on Build GRCh38
rs934764560 Chr.19: 3810749-3810749 on Build GRCh38
rs372091581 Chr.19: 3810728-3810728 on Build GRCh38
rs745888046 Chr.19: 3810741-3810741 on Build GRCh38
rs979019482 Chr.19: 3810893-3810893 on Build GRCh38
rs755697761 Chr.19: 3810692-3810692 on Build GRCh38
rs529162303 Chr.19: 3810772-3810772 on Build GRCh38
rs770983363 Chr.19: 3810738-3810738 on Build GRCh38
rs765883671 Chr.19: 3810812-3810812 on Build GRCh38
rs895529745 Chr.19: 3810840-3810840 on Build GRCh38
rs1023226797 Chr.19: 3810700-3810700 on Build GRCh38
rs147071866 Chr.19: 3810733-3810733 on Build GRCh38
rs1006292803 Chr.19: 3810853-3810853 on Build GRCh38
rs553958404 Chr.19: 3810898-3810898 on Build GRCh38

View Full Product Details