Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 57933732-57933993 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF418
Entrez Gene ID
147686
Gene Name
zinc finger protein 418
Gene Aliases
-
UniGene
Hs.660728
Gene Chromosome Location
Chr.19: 57921882-57935404 on Build GRCh38
Transcripts
NM_001317027.1 NM_001317028.1 NM_001317029.1 NM_001317030.1 NM_133460.2 XM_017026304.1 XM_017026305.1 XM_017026306.1 XM_017026307.1 XM_017026308.1 XM_017026309.1 XM_017026310.1 XM_017026311.1 XM_017026312.1 XM_017026313.1 XM_017026314.1

SNP Detail

SNP ID Chromosome Location
rs373490496 Chr.19: 57933860-57933860 on Build GRCh38
rs773431434 Chr.19: 57933823-57933823 on Build GRCh38
rs775081767 Chr.19: 57933834-57933834 on Build GRCh38
rs915679825 Chr.19: 57933896-57933896 on Build GRCh38
rs776562113 Chr.19: 57933943-57933943 on Build GRCh38
rs568761380 Chr.19: 57933910-57933910 on Build GRCh38
rs574666968 Chr.19: 57933955-57933955 on Build GRCh38
rs773652883 Chr.19: 57933930-57933930 on Build GRCh38
rs889552742 Chr.19: 57933798-57933798 on Build GRCh38
rs748415731 Chr.19: 57933815-57933815 on Build GRCh38
rs751325903 Chr.19: 57933767-57933767 on Build GRCh38
rs748332159 Chr.19: 57933862-57933862 on Build GRCh38
rs763420864 Chr.19: 57933939-57933939 on Build GRCh38
rs755311563 Chr.19: 57933858-57933858 on Build GRCh38
rs369928455 Chr.19: 57933833-57933833 on Build GRCh38
rs896858003 Chr.19: 57933763-57933763 on Build GRCh38
rs558490563 Chr.19: 57933891-57933891 on Build GRCh38
rs930973645 Chr.19: 57933782-57933782 on Build GRCh38
rs1007350554 Chr.19: 57933962-57933962 on Build GRCh38
rs200704288 Chr.19: 57933801-57933801 on Build GRCh38
rs375546575 Chr.19: 57933792-57933792 on Build GRCh38
rs1020231617 Chr.19: 57933820-57933820 on Build GRCh38
rs143548354 Chr.19: 57933940-57933940 on Build GRCh38
rs999776222 Chr.19: 57933824-57933824 on Build GRCh38
rs1039739581 Chr.19: 57933963-57933963 on Build GRCh38
rs765237067 Chr.19: 57933832-57933832 on Build GRCh38
rs566611521 Chr.19: 57933825-57933825 on Build GRCh38
rs928497749 Chr.19: 57933924-57933924 on Build GRCh38
rs762805008 Chr.19: 57933840-57933840 on Build GRCh38
rs192162922 Chr.19: 57933857-57933857 on Build GRCh38
rs749664692 Chr.19: 57933806-57933806 on Build GRCh38
rs957999444 Chr.19: 57933828-57933828 on Build GRCh38
rs768533017 Chr.19: 57933819-57933819 on Build GRCh38
rs184244171 Chr.19: 57933942-57933942 on Build GRCh38
rs972609609 Chr.19: 57933931-57933931 on Build GRCh38
rs755800766 Chr.19: 57933864-57933867 on Build GRCh38
rs1030313694 Chr.19: 57933771-57933771 on Build GRCh38
rs1048105934 Chr.19: 57933948-57933948 on Build GRCh38
rs751223861 Chr.19: 57933844-57933844 on Build GRCh38
rs892938023 Chr.19: 57933960-57933960 on Build GRCh38
rs770395302 Chr.19: 57933784-57933784 on Build GRCh38
rs1054211264 Chr.19: 57933956-57933956 on Build GRCh38
rs373501702 Chr.19: 57933855-57933855 on Build GRCh38
rs1007030426 Chr.19: 57933803-57933803 on Build GRCh38
rs979939202 Chr.19: 57933917-57933917 on Build GRCh38
rs759021761 Chr.19: 57933874-57933875 on Build GRCh38
rs112540410 Chr.19: 57933786-57933786 on Build GRCh38
rs746319105 Chr.19: 57933796-57933796 on Build GRCh38
rs774541058 Chr.19: 57933810-57933810 on Build GRCh38
rs757000433 Chr.19: 57933850-57933850 on Build GRCh38
rs949725245 Chr.19: 57933906-57933906 on Build GRCh38
rs772307561 Chr.19: 57933865-57933865 on Build GRCh38
rs780603149 Chr.19: 57933856-57933856 on Build GRCh38

View Full Product Details