Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 68717964-68718225 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STARD8
Entrez Gene ID
9754
Gene Name
StAR related lipid transfer domain containing 8
Gene Aliases
ARHGAP38, DLC3, STARTGAP3
UniGene
Hs.95140
Gene Chromosome Location
Chr.X: 68647669-68725842 on Build GRCh38
Transcripts
NM_001142503.2 NM_001142504.2 NM_014725.4 XM_005262314.4 XM_005262315.3 XM_011531069.2 XM_011531070.2

SNP Detail

SNP ID Chromosome Location
rs61743089 Chr.X: 68718198-68718198 on Build GRCh38
rs763720132 Chr.X: 68718024-68718024 on Build GRCh38
rs55962426 Chr.X: 68718134-68718134 on Build GRCh38
rs147571370 Chr.X: 68718078-68718078 on Build GRCh38
rs776432270 Chr.X: 68718051-68718051 on Build GRCh38
rs771544394 Chr.X: 68718103-68718103 on Build GRCh38
rs138099266 Chr.X: 68718137-68718137 on Build GRCh38
rs756072446 Chr.X: 68718072-68718072 on Build GRCh38
rs937537328 Chr.X: 68718133-68718133 on Build GRCh38
rs374827815 Chr.X: 68718181-68718181 on Build GRCh38
rs1041950455 Chr.X: 68718149-68718149 on Build GRCh38
rs1033297608 Chr.X: 68718028-68718028 on Build GRCh38
rs1000365227 Chr.X: 68717997-68717997 on Build GRCh38
rs747375700 Chr.X: 68718099-68718099 on Build GRCh38
rs191372880 Chr.X: 68718013-68718013 on Build GRCh38
rs777230336 Chr.X: 68718115-68718115 on Build GRCh38
rs965910723 Chr.X: 68718058-68718058 on Build GRCh38
rs145453283 Chr.X: 68718136-68718136 on Build GRCh38
rs149053296 Chr.X: 68718081-68718081 on Build GRCh38
rs370461954 Chr.X: 68718180-68718180 on Build GRCh38
rs766140204 Chr.X: 68718016-68718016 on Build GRCh38
rs761185526 Chr.X: 68717989-68717989 on Build GRCh38
rs755949443 Chr.X: 68718144-68718144 on Build GRCh38
rs201447670 Chr.X: 68718068-68718068 on Build GRCh38
rs1011940458 Chr.X: 68718032-68718032 on Build GRCh38
rs1007778292 Chr.X: 68718019-68718019 on Build GRCh38
rs768659300 Chr.X: 68718117-68718117 on Build GRCh38
rs756868218 Chr.X: 68718045-68718045 on Build GRCh38
rs767049286 Chr.X: 68718000-68718000 on Build GRCh38
rs143103482 Chr.X: 68718096-68718096 on Build GRCh38
rs748333737 Chr.X: 68718088-68718088 on Build GRCh38
rs760151959 Chr.X: 68718116-68718116 on Build GRCh38
rs754914170 Chr.X: 68718172-68718172 on Build GRCh38
rs371143044 Chr.X: 68718030-68718030 on Build GRCh38
rs867092898 Chr.X: 68718191-68718191 on Build GRCh38
rs202230245 Chr.X: 68718082-68718082 on Build GRCh38
rs372522625 Chr.X: 68718073-68718073 on Build GRCh38
rs766179834 Chr.X: 68718163-68718163 on Build GRCh38
rs758639579 Chr.X: 68718189-68718189 on Build GRCh38
rs773750347 Chr.X: 68718131-68718131 on Build GRCh38
rs151192738 Chr.X: 68718097-68718097 on Build GRCh38
rs746018366 Chr.X: 68718036-68718036 on Build GRCh38
rs888984614 Chr.X: 68717992-68717992 on Build GRCh38
rs763636210 Chr.X: 68718129-68718129 on Build GRCh38

View Full Product Details