Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 17149365-17149626 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CECR5
Entrez Gene ID
27440
Gene Name
cat eye syndrome chromosome region, candidate 5
Gene Aliases
-
UniGene
Hs.26890
Gene Chromosome Location
Chr.22: 17137520-17165287 on Build GRCh38
Transcripts
NM_017829.5 NM_033070.2 XM_005261236.3 XM_011546126.2 XM_011546127.1 XM_017028781.1 XM_017028782.1 XM_017028783.1 XM_017028784.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5017976 c.181G>T p.A61S Chr.22: 17149601-17149601 on Build GRCh38
5197731 c.327C>T p.C109C Chr.22: 17149545-17149545 on Build GRCh38
4418902 c.186G>T p.G62G Chr.22: 17149596-17149596 on Build GRCh38
5017975 c.271G>T p.A91S Chr.22: 17149601-17149601 on Build GRCh38
6626030 c.250C>T p.P84S Chr.22: 17149622-17149622 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148993169 Chr.22: 17149592-17149592 on Build GRCh38
rs71326301 Chr.22: 17149503-17149503 on Build GRCh38
rs1027494137 Chr.22: 17149476-17149476 on Build GRCh38
rs773431442 Chr.22: 17149599-17149599 on Build GRCh38
rs1003442679 Chr.22: 17149432-17149432 on Build GRCh38
rs375543947 Chr.22: 17149521-17149521 on Build GRCh38
rs962223350 Chr.22: 17149460-17149460 on Build GRCh38
rs780792685 Chr.22: 17149559-17149559 on Build GRCh38
rs754936342 Chr.22: 17149559-17149560 on Build GRCh38
rs761547585 Chr.22: 17149494-17149494 on Build GRCh38
rs989217128 Chr.22: 17149595-17149595 on Build GRCh38
rs2286956 Chr.22: 17149488-17149488 on Build GRCh38
rs149869956 Chr.22: 17149593-17149593 on Build GRCh38
rs965608058 Chr.22: 17149528-17149528 on Build GRCh38
rs773630151 Chr.22: 17149498-17149498 on Build GRCh38
rs374271236 Chr.22: 17149587-17149587 on Build GRCh38
rs780008471 Chr.22: 17149584-17149584 on Build GRCh38
rs1034859 Chr.22: 17149596-17149596 on Build GRCh38
rs1034858 Chr.22: 17149499-17149499 on Build GRCh38
rs753799289 Chr.22: 17149518-17149518 on Build GRCh38
rs200633672 Chr.22: 17149569-17149569 on Build GRCh38
rs200337729 Chr.22: 17149544-17149544 on Build GRCh38
rs1014700063 Chr.22: 17149439-17149439 on Build GRCh38
rs977036315 Chr.22: 17149550-17149550 on Build GRCh38
rs759741448 Chr.22: 17149520-17149520 on Build GRCh38
rs756043243 Chr.22: 17149571-17149571 on Build GRCh38
rs773921995 Chr.22: 17149493-17149493 on Build GRCh38
rs772283635 Chr.22: 17149496-17149496 on Build GRCh38
rs201897777 Chr.22: 17149511-17149511 on Build GRCh38
rs906440057 Chr.22: 17149424-17149424 on Build GRCh38
rs758655588 Chr.22: 17149546-17149546 on Build GRCh38
rs761184851 Chr.22: 17149606-17149606 on Build GRCh38
rs748771648 Chr.22: 17149585-17149585 on Build GRCh38
rs994227716 Chr.22: 17149475-17149475 on Build GRCh38
rs1044937601 Chr.22: 17149394-17149394 on Build GRCh38

View Full Product Details