Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 132678905-132679172 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTF3C4
Entrez Gene ID
9329
Gene Name
general transcription factor IIIC subunit 4
Gene Aliases
KAT12, TF3C-delta, TFIII90, TFIIIC290, TFIIIC90, TFIIICDELTA
UniGene
Hs.22302
Gene Chromosome Location
Chr.9: 132670035-132694955 on Build GRCh38
Transcripts
NM_012204.3 NR_133925.1 XM_017015317.1 XM_017015318.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4952623 c.1413G>T p.V471V Chr.9: 132679032-132679032 on Build GRCh38
4516037 c.1530_1531CC>TT p.Q511* Chr.9: 132679149-132679150 on Build GRCh38
5716097 c.1360T>A p.F454I Chr.9: 132678979-132678979 on Build GRCh38
1196570 c.1468G>A p.A490T Chr.9: 132679087-132679087 on Build GRCh38
1756013 c.1362C>T p.F454F Chr.9: 132678981-132678981 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762482369 Chr.9: 132679032-132679032 on Build GRCh38
rs767741207 Chr.9: 132679063-132679063 on Build GRCh38
rs780605582 Chr.9: 132679099-132679099 on Build GRCh38
rs763672314 Chr.9: 132679038-132679038 on Build GRCh38
rs759521618 Chr.9: 132678958-132678958 on Build GRCh38
rs779074814 Chr.9: 132679125-132679125 on Build GRCh38
rs900835851 Chr.9: 132678976-132678976 on Build GRCh38
rs748419455 Chr.9: 132678944-132678944 on Build GRCh38
rs144756359 Chr.9: 132678951-132678951 on Build GRCh38
rs754958605 Chr.9: 132679116-132679116 on Build GRCh38
rs751123154 Chr.9: 132679049-132679049 on Build GRCh38
rs770585023 Chr.9: 132678946-132678946 on Build GRCh38
rs917849815 Chr.9: 132679079-132679079 on Build GRCh38
rs147481161 Chr.9: 132679053-132679053 on Build GRCh38
rs142926692 Chr.9: 132679133-132679133 on Build GRCh38
rs749801864 Chr.9: 132679114-132679114 on Build GRCh38
rs765321046 Chr.9: 132678987-132678987 on Build GRCh38
rs978774425 Chr.9: 132679010-132679010 on Build GRCh38
rs956649378 Chr.9: 132679054-132679054 on Build GRCh38
rs775131333 Chr.9: 132678994-132678994 on Build GRCh38
rs375549318 Chr.9: 132678966-132678966 on Build GRCh38
rs148572797 Chr.9: 132679080-132679080 on Build GRCh38
rs755530765 Chr.9: 132679075-132679075 on Build GRCh38
rs926331620 Chr.9: 132679122-132679122 on Build GRCh38
rs201142583 Chr.9: 132679074-132679074 on Build GRCh38

View Full Product Details