Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 109643928-109644159 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSL4
Entrez Gene ID
2182
Gene Name
acyl-CoA synthetase long-chain family member 4
Gene Aliases
ACS4, FACL4, LACS4, MRX63, MRX68
UniGene
Hs.268785
Gene Chromosome Location
Chr.X: 109641330-109733403 on Build GRCh38
Transcripts
NM_001318509.1 NM_001318510.1 NM_004458.2 NM_022977.2 XM_005262109.2 XM_006724635.2 XM_011530888.2 XM_011530889.2

SNP Detail

SNP ID Chromosome Location
rs760480845 Chr.X: 109644019-109644019 on Build GRCh38
rs184672678 Chr.X: 109643992-109643992 on Build GRCh38
rs755004249 Chr.X: 109644128-109644128 on Build GRCh38
rs1056037005 Chr.X: 109643977-109643977 on Build GRCh38
rs752203390 Chr.X: 109644137-109644137 on Build GRCh38
rs769828137 Chr.X: 109644001-109644001 on Build GRCh38
rs756342527 Chr.X: 109644011-109644012 on Build GRCh38
rs777069871 Chr.X: 109643994-109643994 on Build GRCh38
rs764281998 Chr.X: 109644008-109644008 on Build GRCh38
rs150760891 Chr.X: 109644002-109644002 on Build GRCh38
rs766299247 Chr.X: 109644036-109644036 on Build GRCh38
rs973692121 Chr.X: 109643981-109643981 on Build GRCh38
rs898008437 Chr.X: 109644070-109644070 on Build GRCh38
rs763217379 Chr.X: 109644007-109644007 on Build GRCh38
rs764780876 Chr.X: 109644134-109644134 on Build GRCh38
rs376192104 Chr.X: 109644113-109644113 on Build GRCh38
rs765895091 Chr.X: 109643999-109643999 on Build GRCh38
rs1802109 Chr.X: 109643975-109643975 on Build GRCh38
rs987531901 Chr.X: 109644069-109644069 on Build GRCh38
rs773057024 Chr.X: 109644015-109644015 on Build GRCh38

View Full Product Details