Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 165497345-165497582 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPE
Entrez Gene ID
1363
Gene Name
carboxypeptidase E
Gene Aliases
CPH
UniGene
Hs.75360
Gene Chromosome Location
Chr.4: 165378942-165498547 on Build GRCh38
Transcripts
NM_001873.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1052962 c.1369G>T p.E457* Chr.4: 165497548-165497548 on Build GRCh38
7180889 c.1394A>T p.E465V Chr.4: 165497573-165497573 on Build GRCh38
1052963 c.1371A>C p.E457D Chr.4: 165497550-165497550 on Build GRCh38
6050628 c.1342G>C p.E448Q Chr.4: 165497521-165497521 on Build GRCh38
1052961 c.1348G>T p.E450* Chr.4: 165497527-165497527 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs560038598 Chr.4: 165497545-165497545 on Build GRCh38
rs761072769 Chr.4: 165497513-165497513 on Build GRCh38
rs770548812 Chr.4: 165497467-165497467 on Build GRCh38
rs951331026 Chr.4: 165497542-165497542 on Build GRCh38
rs963394508 Chr.4: 165497393-165497393 on Build GRCh38
rs112020947 Chr.4: 165497481-165497481 on Build GRCh38
rs772486883 Chr.4: 165497503-165497503 on Build GRCh38
rs867354434 Chr.4: 165497412-165497412 on Build GRCh38
rs539600557 Chr.4: 165497512-165497512 on Build GRCh38
rs570592172 Chr.4: 165497389-165497389 on Build GRCh38
rs917080769 Chr.4: 165497457-165497457 on Build GRCh38
rs765444839 Chr.4: 165497556-165497556 on Build GRCh38
rs565165400 Chr.4: 165497374-165497374 on Build GRCh38
rs28465223 Chr.4: 165497424-165497424 on Build GRCh38
rs759037468 Chr.4: 165497490-165497490 on Build GRCh38
rs750495991 Chr.4: 165497557-165497557 on Build GRCh38
rs958571595 Chr.4: 165497409-165497409 on Build GRCh38
rs764427251 Chr.4: 165497516-165497516 on Build GRCh38
rs748936170 Chr.4: 165497462-165497462 on Build GRCh38
rs995311140 Chr.4: 165497420-165497420 on Build GRCh38
rs767414770 Chr.4: 165497541-165497541 on Build GRCh38
rs1008095617 Chr.4: 165497381-165497381 on Build GRCh38

View Full Product Details