Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 100916966-100917205 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLF2
Entrez Gene ID
55719
Gene Name
SMC5-SMC6 complex localization factor 2
Gene Aliases
C10orf6, FAM178A
UniGene
Hs.447458
Gene Chromosome Location
Chr.10: 100910230-100965134 on Build GRCh38
Transcripts
NM_001136123.1 NM_018121.3 XM_005269965.2 XM_011539944.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4944164 c.675C>T p.H225H Chr.10: 100917060-100917060 on Build GRCh38
673541 c.620A>T p.D207V Chr.10: 100917005-100917005 on Build GRCh38
2058136 c.739C>T p.R247* Chr.10: 100917124-100917124 on Build GRCh38
7367170 c.641G>A p.R214K Chr.10: 100917026-100917026 on Build GRCh38
415201 c.756G>C p.L252F Chr.10: 100917141-100917141 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768092411 Chr.10: 100917159-100917159 on Build GRCh38
rs770511394 Chr.10: 100916988-100916989 on Build GRCh38
rs772081455 Chr.10: 100917007-100917007 on Build GRCh38
rs756286289 Chr.10: 100917129-100917129 on Build GRCh38
rs372105225 Chr.10: 100917115-100917115 on Build GRCh38
rs764665334 Chr.10: 100917054-100917054 on Build GRCh38
rs113874914 Chr.10: 100917175-100917175 on Build GRCh38
rs909412454 Chr.10: 100917091-100917091 on Build GRCh38
rs775944288 Chr.10: 100917035-100917035 on Build GRCh38
rs562339204 Chr.10: 100917090-100917090 on Build GRCh38
rs376364235 Chr.10: 100917073-100917073 on Build GRCh38
rs143748124 Chr.10: 100917078-100917078 on Build GRCh38
rs763185656 Chr.10: 100917050-100917050 on Build GRCh38
rs771056858 Chr.10: 100917023-100917023 on Build GRCh38
rs775187506 Chr.10: 100917029-100917029 on Build GRCh38
rs753432952 Chr.10: 100917161-100917161 on Build GRCh38
rs147132029 Chr.10: 100917065-100917065 on Build GRCh38
rs770084590 Chr.10: 100917096-100917096 on Build GRCh38
rs747851722 Chr.10: 100917004-100917004 on Build GRCh38
rs778393400 Chr.10: 100917066-100917066 on Build GRCh38
rs139059266 Chr.10: 100917026-100917026 on Build GRCh38
rs774768767 Chr.10: 100917150-100917150 on Build GRCh38
rs140623866 Chr.10: 100917105-100917105 on Build GRCh38
rs775505573 Chr.10: 100917102-100917102 on Build GRCh38
rs770823757 Chr.10: 100917084-100917084 on Build GRCh38
rs747981951 Chr.10: 100917069-100917069 on Build GRCh38
rs762464041 Chr.10: 100917157-100917157 on Build GRCh38
rs772958504 Chr.10: 100917014-100917014 on Build GRCh38
rs532419131 Chr.10: 100917030-100917030 on Build GRCh38
rs760823060 Chr.10: 100917021-100917021 on Build GRCh38
rs145002981 Chr.10: 100917057-100917057 on Build GRCh38
rs757821791 Chr.10: 100917058-100917058 on Build GRCh38
rs745898055 Chr.10: 100917093-100917093 on Build GRCh38
rs764420815 Chr.10: 100917148-100917148 on Build GRCh38
rs765421964 Chr.10: 100917032-100917032 on Build GRCh38
rs777612233 Chr.10: 100917077-100917077 on Build GRCh38
rs778821263 Chr.10: 100917002-100917002 on Build GRCh38

View Full Product Details