Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 92569660-92569899 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM133B
Entrez Gene ID
257415
Gene Name
family with sequence similarity 133 member B
Gene Aliases
-
UniGene
Hs.18564
Gene Chromosome Location
Chr.7: 92560758-92590394 on Build GRCh38
Transcripts
NM_001040057.2 NM_001288584.1 NM_152789.3 NR_109929.1 XM_011516004.2 XM_011516005.2

SNP Detail

SNP ID Chromosome Location
rs3841401 Chr.7: 92569702-92569702 on Build GRCh38
rs949673513 Chr.7: 92569717-92569717 on Build GRCh38
rs942166812 Chr.7: 92569860-92569860 on Build GRCh38
rs1047942321 Chr.7: 92569727-92569727 on Build GRCh38
rs1050234317 Chr.7: 92569842-92569842 on Build GRCh38
rs771282560 Chr.7: 92569775-92569778 on Build GRCh38
rs759080347 Chr.7: 92569775-92569775 on Build GRCh38
rs931926870 Chr.7: 92569751-92569751 on Build GRCh38
rs775225460 Chr.7: 92569830-92569830 on Build GRCh38
rs760409424 Chr.7: 92569862-92569862 on Build GRCh38
rs397841682 Chr.7: 92569707-92569707 on Build GRCh38
rs543096025 Chr.7: 92569814-92569814 on Build GRCh38

View Full Product Details