Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 161294216-161294463 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRB2
Entrez Gene ID
2561
Gene Name
gamma-aminobutyric acid type A receptor beta2 subunit
Gene Aliases
-
UniGene
Hs.303527
Gene Chromosome Location
Chr.5: 161288429-161548124 on Build GRCh38
Transcripts
NM_000813.2 NM_021911.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4790556 c.1274G>T p.G425V Chr.5: 161294346-161294346 on Build GRCh38
736936 c.1195T>C p.S399P Chr.5: 161294311-161294311 on Build GRCh38
4589809 c.1125_1126insA p.E376fs*6 Chr.5: 161294380-161294381 on Build GRCh38
736935 c.1309T>C p.S437P Chr.5: 161294311-161294311 on Build GRCh38
387611 c.1149G>A p.V383V Chr.5: 161294357-161294357 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs981300732 Chr.5: 161294272-161294272 on Build GRCh38
rs777255206 Chr.5: 161294326-161294326 on Build GRCh38
rs867485613 Chr.5: 161294375-161294375 on Build GRCh38
rs146425610 Chr.5: 161294393-161294393 on Build GRCh38
rs947062030 Chr.5: 161294362-161294362 on Build GRCh38
rs371728438 Chr.5: 161294262-161294262 on Build GRCh38
rs201342925 Chr.5: 161294243-161294243 on Build GRCh38
rs758133298 Chr.5: 161294252-161294252 on Build GRCh38
rs866403685 Chr.5: 161294380-161294380 on Build GRCh38
rs761178093 Chr.5: 161294244-161294244 on Build GRCh38
rs75580767 Chr.5: 161294347-161294347 on Build GRCh38
rs765952235 Chr.5: 161294341-161294341 on Build GRCh38
rs770274870 Chr.5: 161294408-161294408 on Build GRCh38
rs778847152 Chr.5: 161294371-161294371 on Build GRCh38
rs746075634 Chr.5: 161294304-161294304 on Build GRCh38
rs773594705 Chr.5: 161294414-161294414 on Build GRCh38
rs781071477 Chr.5: 161294286-161294286 on Build GRCh38
rs952831120 Chr.5: 161294238-161294238 on Build GRCh38
rs750901071 Chr.5: 161294410-161294410 on Build GRCh38
rs772398405 Chr.5: 161294310-161294310 on Build GRCh38
rs750461505 Chr.5: 161294388-161294388 on Build GRCh38
rs770453629 Chr.5: 161294420-161294420 on Build GRCh38
rs78260275 Chr.5: 161294353-161294353 on Build GRCh38
rs982764804 Chr.5: 161294242-161294242 on Build GRCh38
rs779091017 Chr.5: 161294295-161294295 on Build GRCh38
rs762487283 Chr.5: 161294417-161294417 on Build GRCh38
rs200591388 Chr.5: 161294398-161294398 on Build GRCh38
rs368394878 Chr.5: 161294281-161294281 on Build GRCh38
rs769164805 Chr.5: 161294322-161294322 on Build GRCh38
rs773974056 Chr.5: 161294431-161294431 on Build GRCh38
rs61735410 Chr.5: 161294271-161294271 on Build GRCh38
rs80241240 Chr.5: 161294247-161294247 on Build GRCh38
rs762419466 Chr.5: 161294327-161294327 on Build GRCh38
rs772833467 Chr.5: 161294330-161294330 on Build GRCh38
rs762381690 Chr.5: 161294340-161294340 on Build GRCh38
rs759226981 Chr.5: 161294434-161294434 on Build GRCh38
rs142424271 Chr.5: 161294407-161294407 on Build GRCh38
rs78789939 Chr.5: 161294416-161294416 on Build GRCh38
rs751246871 Chr.5: 161294343-161294343 on Build GRCh38
rs914459006 Chr.5: 161294361-161294361 on Build GRCh38
rs758579755 Chr.5: 161294390-161294390 on Build GRCh38
rs752546514 Chr.5: 161294363-161294363 on Build GRCh38
rs756144528 Chr.5: 161294368-161294368 on Build GRCh38
rs945039011 Chr.5: 161294367-161294367 on Build GRCh38
rs747335724 Chr.5: 161294315-161294315 on Build GRCh38
rs2229944 Chr.5: 161294312-161294312 on Build GRCh38
rs764850270 Chr.5: 161294246-161294246 on Build GRCh38
rs148772321 Chr.5: 161294392-161294392 on Build GRCh38
rs536960677 Chr.5: 161294237-161294237 on Build GRCh38
rs748111065 Chr.5: 161294289-161294289 on Build GRCh38
rs778879790 Chr.5: 161294360-161294360 on Build GRCh38
rs764645599 Chr.5: 161294329-161294329 on Build GRCh38

View Full Product Details