Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 152308478-152308719 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLG
Entrez Gene ID
2312
Gene Name
filaggrin
Gene Aliases
ATOD2
UniGene
Hs.654510
Gene Chromosome Location
Chr.1: 152302175-152325203 on Build GRCh38
Transcripts
NM_002016.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1667965 c.6357T>G p.Y2119* Chr.1: 152308529-152308529 on Build GRCh38
6664442 c.6183A>G p.A2061A Chr.1: 152308703-152308703 on Build GRCh38
1724902 c.6327C>A p.P2109P Chr.1: 152308559-152308559 on Build GRCh38
5460954 c.6190A>C p.K2064Q Chr.1: 152308696-152308696 on Build GRCh38
3704992 c.6368A>G p.Q2123R Chr.1: 152308518-152308518 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753198584 Chr.1: 152308628-152308628 on Build GRCh38
rs754760489 Chr.1: 152308502-152308502 on Build GRCh38
rs142063898 Chr.1: 152308582-152308582 on Build GRCh38
rs749273002 Chr.1: 152308699-152308699 on Build GRCh38
rs920315793 Chr.1: 152308673-152308673 on Build GRCh38
rs746024413 Chr.1: 152308636-152308636 on Build GRCh38
rs775399724 Chr.1: 152308564-152308564 on Build GRCh38
rs767229540 Chr.1: 152308674-152308674 on Build GRCh38
rs771088791 Chr.1: 152308632-152308632 on Build GRCh38
rs753763512 Chr.1: 152308573-152308573 on Build GRCh38
rs573866396 Chr.1: 152308669-152308669 on Build GRCh38
rs776681299 Chr.1: 152308529-152308529 on Build GRCh38
rs553906156 Chr.1: 152308616-152308616 on Build GRCh38
rs774954937 Chr.1: 152308675-152308675 on Build GRCh38
rs374988997 Chr.1: 152308689-152308689 on Build GRCh38
rs72477392 Chr.1: 152308577-152308577 on Build GRCh38
rs755340161 Chr.1: 152308626-152308626 on Build GRCh38
rs565362443 Chr.1: 152308506-152308506 on Build GRCh38
rs760572823 Chr.1: 152308605-152308605 on Build GRCh38
rs201446270 Chr.1: 152308650-152308650 on Build GRCh38
rs151189270 Chr.1: 152308657-152308657 on Build GRCh38
rs780393149 Chr.1: 152308552-152308552 on Build GRCh38
rs769415669 Chr.1: 152308667-152308667 on Build GRCh38
rs367724692 Chr.1: 152308670-152308670 on Build GRCh38
rs147095782 Chr.1: 152308668-152308668 on Build GRCh38
rs759263160 Chr.1: 152308533-152308533 on Build GRCh38
rs761116243 Chr.1: 152308676-152308676 on Build GRCh38
rs147145234 Chr.1: 152308647-152308647 on Build GRCh38
rs761082529 Chr.1: 152308629-152308629 on Build GRCh38
rs781672763 Chr.1: 152308581-152308581 on Build GRCh38
rs751278554 Chr.1: 152308501-152308501 on Build GRCh38
rs769741410 Chr.1: 152308642-152308642 on Build GRCh38
rs776441293 Chr.1: 152308609-152308609 on Build GRCh38
rs763927334 Chr.1: 152308607-152308607 on Build GRCh38
rs756735418 Chr.1: 152308574-152308574 on Build GRCh38
rs766040205 Chr.1: 152308621-152308621 on Build GRCh38
rs765461482 Chr.1: 152308687-152308687 on Build GRCh38
rs375444567 Chr.1: 152308613-152308613 on Build GRCh38
rs7522925 Chr.1: 152308563-152308563 on Build GRCh38
rs74129455 Chr.1: 152308696-152308696 on Build GRCh38
rs749995949 Chr.1: 152308641-152308642 on Build GRCh38
rs142067855 Chr.1: 152308541-152308541 on Build GRCh38
rs772972657 Chr.1: 152308645-152308645 on Build GRCh38
rs774677530 Chr.1: 152308601-152308601 on Build GRCh38
rs201317833 Chr.1: 152308570-152308570 on Build GRCh38
rs756936662 Chr.1: 152308600-152308601 on Build GRCh38
rs199564792 Chr.1: 152308653-152308653 on Build GRCh38
rs779378312 Chr.1: 152308518-152308518 on Build GRCh38
rs751152856 Chr.1: 152308622-152308622 on Build GRCh38
rs763227797 Chr.1: 152308562-152308562 on Build GRCh38
rs772316804 Chr.1: 152308664-152308664 on Build GRCh38
rs749003404 Chr.1: 152308512-152308512 on Build GRCh38
rs200140094 Chr.1: 152308511-152308511 on Build GRCh38
rs368465259 Chr.1: 152308540-152308540 on Build GRCh38
rs769130811 Chr.1: 152308555-152308555 on Build GRCh38
rs769827566 Chr.1: 152308587-152308587 on Build GRCh38
rs148799492 Chr.1: 152308637-152308637 on Build GRCh38
rs141120727 Chr.1: 152308666-152308666 on Build GRCh38
rs149955442 Chr.1: 152308661-152308661 on Build GRCh38
rs7512554 Chr.1: 152308532-152308532 on Build GRCh38
rs199750855 Chr.1: 152308567-152308567 on Build GRCh38
rs768793175 Chr.1: 152308526-152308526 on Build GRCh38
rs777080413 Chr.1: 152308556-152308556 on Build GRCh38
rs756000944 Chr.1: 152308694-152308694 on Build GRCh38
rs749518907 Chr.1: 152308593-152308593 on Build GRCh38
rs201739837 Chr.1: 152308579-152308579 on Build GRCh38
rs760732380 Chr.1: 152308566-152308566 on Build GRCh38
rs752623727 Chr.1: 152308505-152308505 on Build GRCh38
rs143616671 Chr.1: 152308551-152308551 on Build GRCh38
rs201815027 Chr.1: 152308539-152308539 on Build GRCh38
rs755684943 Chr.1: 152308508-152308508 on Build GRCh38
rs538619112 Chr.1: 152308561-152308561 on Build GRCh38
rs761806799 Chr.1: 152308612-152308612 on Build GRCh38
rs778225885 Chr.1: 152308639-152308639 on Build GRCh38
rs753388685 Chr.1: 152308529-152308531 on Build GRCh38
rs758330162 Chr.1: 152308644-152308645 on Build GRCh38
rs746788657 Chr.1: 152308684-152308684 on Build GRCh38
rs146352383 Chr.1: 152308630-152308630 on Build GRCh38
rs769853707 Chr.1: 152308559-152308559 on Build GRCh38
rs199928720 Chr.1: 152308682-152308682 on Build GRCh38
rs771244281 Chr.1: 152308600-152308600 on Build GRCh38
rs777413192 Chr.1: 152308509-152308509 on Build GRCh38
rs781013387 Chr.1: 152308504-152308504 on Build GRCh38
rs780010597 Chr.1: 152308646-152308646 on Build GRCh38
rs753070694 Chr.1: 152308656-152308656 on Build GRCh38
rs200491819 Chr.1: 152308641-152308641 on Build GRCh38
rs145119684 Chr.1: 152308519-152308519 on Build GRCh38
rs527524627 Chr.1: 152308513-152308513 on Build GRCh38
rs751279387 Chr.1: 152308649-152308649 on Build GRCh38
rs7512553 Chr.1: 152308531-152308531 on Build GRCh38
rs779294204 Chr.1: 152308635-152308635 on Build GRCh38
rs373662277 Chr.1: 152308557-152308557 on Build GRCh38
rs775990772 Chr.1: 152308522-152308522 on Build GRCh38
rs867944258 Chr.1: 152308698-152308698 on Build GRCh38
rs549806447 Chr.1: 152308550-152308550 on Build GRCh38
rs757830053 Chr.1: 152308685-152308685 on Build GRCh38
rs759225149 Chr.1: 152308671-152308671 on Build GRCh38
rs755285154 Chr.1: 152308580-152308580 on Build GRCh38
rs149635053 Chr.1: 152308633-152308633 on Build GRCh38
rs778456715 Chr.1: 152308542-152308542 on Build GRCh38
rs565740303 Chr.1: 152308578-152308578 on Build GRCh38
rs773325239 Chr.1: 152308592-152308592 on Build GRCh38
rs537052355 Chr.1: 152308627-152308627 on Build GRCh38
rs758372218 Chr.1: 152308688-152308688 on Build GRCh38
rs762461321 Chr.1: 152308646-152308646 on Build GRCh38
rs78125326 Chr.1: 152308660-152308660 on Build GRCh38
rs752744646 Chr.1: 152308537-152308537 on Build GRCh38
rs762020950 Chr.1: 152308500-152308500 on Build GRCh38
rs761105257 Chr.1: 152308523-152308523 on Build GRCh38
rs141923137 Chr.1: 152308681-152308681 on Build GRCh38

View Full Product Details