Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 168379563-168379822 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX60L
Entrez Gene ID
91351
Gene Name
DEAD-box helicase 60-like
Gene Aliases
-
UniGene
Hs.535011
Gene Chromosome Location
Chr.4: 168356735-168480514 on Build GRCh38
Transcripts
NM_001012967.2 XM_005263341.4 XM_005263342.2 XM_005263343.2 XM_005263345.4 XM_006714408.2 XM_011532402.2 XM_011532403.2 XM_011532404.1 XM_011532405.1 XM_011532406.1 XM_011532409.2 XM_017008829.1 XM_017008830.1 XM_017008831.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4793162 c.4206G>A p.Q1402Q Chr.4: 168379741-168379741 on Build GRCh38
5730079 c.4146G>A p.L1382L Chr.4: 168379801-168379801 on Build GRCh38
4135489 c.4216A>T p.K1406* Chr.4: 168379731-168379731 on Build GRCh38
6298590 c.4159C>T p.R1387* Chr.4: 168379788-168379788 on Build GRCh38
260385 c.4173G>A p.E1391E Chr.4: 168379774-168379774 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs922577860 Chr.4: 168379623-168379623 on Build GRCh38
rs147726463 Chr.4: 168379667-168379667 on Build GRCh38
rs532154150 Chr.4: 168379646-168379646 on Build GRCh38
rs1042687447 Chr.4: 168379614-168379614 on Build GRCh38
rs769417927 Chr.4: 168379723-168379723 on Build GRCh38
rs774323165 Chr.4: 168379778-168379778 on Build GRCh38
rs1026259779 Chr.4: 168379729-168379729 on Build GRCh38
rs746729525 Chr.4: 168379654-168379654 on Build GRCh38
rs993803841 Chr.4: 168379718-168379718 on Build GRCh38
rs1022343638 Chr.4: 168379761-168379762 on Build GRCh38
rs61740720 Chr.4: 168379787-168379787 on Build GRCh38
rs780667983 Chr.4: 168379704-168379704 on Build GRCh38
rs899036693 Chr.4: 168379693-168379693 on Build GRCh38
rs770827594 Chr.4: 168379773-168379773 on Build GRCh38
rs907138620 Chr.4: 168379681-168379681 on Build GRCh38
rs560176588 Chr.4: 168379702-168379702 on Build GRCh38
rs971065889 Chr.4: 168379755-168379755 on Build GRCh38
rs760626682 Chr.4: 168379779-168379779 on Build GRCh38
rs79652210 Chr.4: 168379624-168379624 on Build GRCh38
rs945613617 Chr.4: 168379611-168379611 on Build GRCh38
rs1041098855 Chr.4: 168379697-168379697 on Build GRCh38
rs772980074 Chr.4: 168379736-168379736 on Build GRCh38
rs201759136 Chr.4: 168379737-168379737 on Build GRCh38
rs764269076 Chr.4: 168379782-168379782 on Build GRCh38
rs780745188 Chr.4: 168379684-168379684 on Build GRCh38
rs1046903481 Chr.4: 168379677-168379677 on Build GRCh38
rs1035459799 Chr.4: 168379700-168379700 on Build GRCh38
rs543303109 Chr.4: 168379630-168379630 on Build GRCh38
rs746582222 Chr.4: 168379729-168379730 on Build GRCh38
rs765767331 Chr.4: 168379663-168379663 on Build GRCh38
rs373439025 Chr.4: 168379717-168379717 on Build GRCh38
rs564882318 Chr.4: 168379631-168379631 on Build GRCh38
rs369050206 Chr.4: 168379683-168379683 on Build GRCh38
rs79153777 Chr.4: 168379600-168379600 on Build GRCh38
rs377435405 Chr.4: 168379725-168379725 on Build GRCh38
rs746493457 Chr.4: 168379690-168379690 on Build GRCh38
rs978248221 Chr.4: 168379794-168379794 on Build GRCh38
rs373131010 Chr.4: 168379788-168379788 on Build GRCh38
rs551734786 Chr.4: 168379741-168379741 on Build GRCh38

View Full Product Details