Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 123383772-123383999 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DENND1A
Entrez Gene ID
57706
Gene Name
DENN domain containing 1A
Gene Aliases
FAM31A, KIAA1608
UniGene
Hs.744995
Gene Chromosome Location
Chr.9: 123379654-123930158 on Build GRCh38
Transcripts
NM_020946.1 XM_005252109.2 XM_005252111.4 XM_006717195.3 XM_011518882.2 XM_011518883.2 XM_011518884.2 XM_011518885.2 XM_011518886.2 XM_017014947.1 XM_017014948.1 XM_017014949.1 XM_017014950.1 XM_017014951.1 XM_017014952.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3324325 c.1615G>A p.A539T Chr.9: 123383876-123383876 on Build GRCh38
4382998 c.1672C>A p.P558T Chr.9: 123383852-123383852 on Build GRCh38
3324323 c.1626C>T p.D542D Chr.9: 123383865-123383865 on Build GRCh38
3664076 c.1624C>T p.R542W Chr.9: 123383900-123383900 on Build GRCh38
1666508 c.1615_1616insCA p.Q539fs*67 Chr.9: 123383908-123383909 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780392876 Chr.9: 123383917-123383918 on Build GRCh38
rs576783467 Chr.9: 123383972-123383972 on Build GRCh38
rs752178995 Chr.9: 123383911-123383911 on Build GRCh38
rs754580261 Chr.9: 123383828-123383828 on Build GRCh38
rs769795344 Chr.9: 123383958-123383958 on Build GRCh38
rs75693594 Chr.9: 123383910-123383910 on Build GRCh38
rs780491508 Chr.9: 123383864-123383864 on Build GRCh38
rs767530779 Chr.9: 123383913-123383918 on Build GRCh38
rs779521644 Chr.9: 123383852-123383852 on Build GRCh38
rs756669110 Chr.9: 123383913-123383914 on Build GRCh38
rs951786561 Chr.9: 123383969-123383969 on Build GRCh38
rs762737817 Chr.9: 123383886-123383886 on Build GRCh38
rs371715901 Chr.9: 123383876-123383876 on Build GRCh38
rs768614573 Chr.9: 123383799-123383799 on Build GRCh38
rs765844102 Chr.9: 123383914-123383914 on Build GRCh38
rs371253977 Chr.9: 123383819-123383819 on Build GRCh38
rs149225844 Chr.9: 123383857-123383857 on Build GRCh38
rs773025069 Chr.9: 123383806-123383806 on Build GRCh38
rs751102366 Chr.9: 123383826-123383826 on Build GRCh38
rs1010017703 Chr.9: 123383935-123383935 on Build GRCh38
rs546579684 Chr.9: 123383836-123383836 on Build GRCh38
rs754134734 Chr.9: 123383931-123383932 on Build GRCh38
rs111273094 Chr.9: 123383918-123383918 on Build GRCh38
rs766154847 Chr.9: 123383900-123383900 on Build GRCh38
rs948085165 Chr.9: 123383897-123383897 on Build GRCh38
rs1000902921 Chr.9: 123383905-123383905 on Build GRCh38
rs974474775 Chr.9: 123383821-123383821 on Build GRCh38
rs199648302 Chr.9: 123383801-123383801 on Build GRCh38
rs987183920 Chr.9: 123383890-123383890 on Build GRCh38
rs146395927 Chr.9: 123383940-123383940 on Build GRCh38
rs755189353 Chr.9: 123383932-123383932 on Build GRCh38
rs368054934 Chr.9: 123383875-123383875 on Build GRCh38
rs145330954 Chr.9: 123383823-123383823 on Build GRCh38
rs965855533 Chr.9: 123383952-123383952 on Build GRCh38
rs146918976 Chr.9: 123383904-123383904 on Build GRCh38
rs773171723 Chr.9: 123383854-123383855 on Build GRCh38
rs375875060 Chr.9: 123383865-123383865 on Build GRCh38
rs2808411 Chr.9: 123383918-123383918 on Build GRCh38
rs755358789 Chr.9: 123383958-123383959 on Build GRCh38
rs760746802 Chr.9: 123383878-123383881 on Build GRCh38
rs748187165 Chr.9: 123383957-123383957 on Build GRCh38
rs1028757296 Chr.9: 123383960-123383960 on Build GRCh38
rs773198671 Chr.9: 123383961-123383961 on Build GRCh38
rs758892707 Chr.9: 123383917-123383917 on Build GRCh38
rs758771557 Chr.9: 123383862-123383862 on Build GRCh38
rs2808410 Chr.9: 123383859-123383859 on Build GRCh38
rs755635673 Chr.9: 123383913-123383913 on Build GRCh38
rs750961169 Chr.9: 123383915-123383915 on Build GRCh38
rs750917810 Chr.9: 123383913-123383915 on Build GRCh38
rs757672795 Chr.9: 123383843-123383843 on Build GRCh38
rs776546729 Chr.9: 123383870-123383870 on Build GRCh38
rs201558847 Chr.9: 123383877-123383877 on Build GRCh38
rs770839373 Chr.9: 123383908-123383909 on Build GRCh38
rs1042337594 Chr.9: 123383947-123383947 on Build GRCh38
rs117892492 Chr.9: 123383868-123383868 on Build GRCh38
rs754417395 Chr.9: 123383837-123383837 on Build GRCh38
rs141135739 Chr.9: 123383874-123383874 on Build GRCh38
rs776689069 Chr.9: 123383800-123383800 on Build GRCh38

View Full Product Details