Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 35660959-35661232 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF39
Entrez Gene ID
84904
Gene Name
Rho guanine nucleotide exchange factor 39
Gene Aliases
C9orf100
UniGene
Hs.534579
Gene Chromosome Location
Chr.9: 35658875-35665281 on Build GRCh38
Transcripts
NM_032818.2 XM_011518057.2 XM_017015225.1 XM_017015227.1 NM_001195200.1 NM_001195201.1 NM_001195217.1 NM_174923.2 XM_005251403.4

Gene Symbol
CCDC107
Entrez Gene ID
203260
Gene Name
coiled-coil domain containing 107
Gene Aliases
PSEC0222
UniGene
Hs.745107
Gene Chromosome Location
Chr.9: 35658282-35661503 on Build GRCh38
Transcripts
NM_032818.2 XM_011518057.2 XM_017015225.1 XM_017015227.1 NM_001195200.1 NM_001195201.1 NM_001195217.1 NM_174923.2 XM_005251403.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1108684 c.777G>C p.K259N Chr.9: 35661112-35661112 on Build GRCh38
3907188 c.749G>A p.R250Q Chr.9: 35661084-35661084 on Build GRCh38
4669661 c.672G>A p.Q224Q Chr.9: 35661007-35661007 on Build GRCh38
6255825 c.684G>T p.E228D Chr.9: 35661019-35661019 on Build GRCh38
5640954 c.839G>C p.S280T Chr.9: 35661174-35661174 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747983010 Chr.9: 35661177-35661177 on Build GRCh38
rs139159879 Chr.9: 35661166-35661166 on Build GRCh38
rs774688219 Chr.9: 35661143-35661143 on Build GRCh38
rs756761846 Chr.9: 35661084-35661084 on Build GRCh38
rs10441686 Chr.9: 35661091-35661091 on Build GRCh38
rs1042864491 Chr.9: 35661052-35661052 on Build GRCh38
rs761267468 Chr.9: 35661153-35661153 on Build GRCh38
rs763119314 Chr.9: 35661133-35661133 on Build GRCh38
rs766678660 Chr.9: 35661067-35661067 on Build GRCh38
rs777404170 Chr.9: 35661103-35661104 on Build GRCh38
rs199818206 Chr.9: 35661031-35661031 on Build GRCh38
rs779926968 Chr.9: 35661098-35661098 on Build GRCh38
rs748224933 Chr.9: 35661123-35661123 on Build GRCh38
rs534657208 Chr.9: 35661001-35661004 on Build GRCh38
rs748763882 Chr.9: 35660997-35660997 on Build GRCh38
rs775450547 Chr.9: 35661036-35661036 on Build GRCh38
rs71521284 Chr.9: 35661017-35661017 on Build GRCh38
rs765492244 Chr.9: 35661061-35661061 on Build GRCh38
rs781163461 Chr.9: 35661175-35661175 on Build GRCh38
rs751950189 Chr.9: 35661070-35661070 on Build GRCh38
rs747003370 Chr.9: 35661115-35661115 on Build GRCh38
rs769798574 Chr.9: 35661178-35661178 on Build GRCh38
rs749430166 Chr.9: 35661185-35661185 on Build GRCh38
rs10441685 Chr.9: 35661060-35661060 on Build GRCh38
rs754860048 Chr.9: 35661174-35661174 on Build GRCh38
rs763277783 Chr.9: 35661065-35661065 on Build GRCh38
rs377274752 Chr.9: 35661148-35661148 on Build GRCh38
rs35921814 Chr.9: 35661010-35661010 on Build GRCh38
rs764613156 Chr.9: 35661159-35661159 on Build GRCh38
rs753387364 Chr.9: 35661083-35661083 on Build GRCh38
rs766836308 Chr.9: 35660983-35660983 on Build GRCh38
rs769809480 Chr.9: 35661125-35661125 on Build GRCh38
rs147165051 Chr.9: 35660994-35660994 on Build GRCh38
rs113536720 Chr.9: 35661097-35661097 on Build GRCh38
rs561977892 Chr.9: 35661119-35661119 on Build GRCh38
rs778499991 Chr.9: 35661009-35661009 on Build GRCh38
rs747714583 Chr.9: 35660991-35660993 on Build GRCh38
rs879048977 Chr.9: 35661078-35661078 on Build GRCh38
rs753194449 Chr.9: 35661149-35661149 on Build GRCh38
rs1339374 Chr.9: 35660993-35660993 on Build GRCh38
rs746904340 Chr.9: 35661056-35661056 on Build GRCh38
rs996429504 Chr.9: 35661201-35661201 on Build GRCh38
rs774600251 Chr.9: 35661198-35661198 on Build GRCh38
rs749959956 Chr.9: 35661161-35661161 on Build GRCh38
rs755422129 Chr.9: 35661071-35661071 on Build GRCh38
rs201575851 Chr.9: 35661066-35661066 on Build GRCh38
rs756924613 Chr.9: 35660998-35660998 on Build GRCh38
rs773343294 Chr.9: 35661064-35661064 on Build GRCh38
rs149945904 Chr.9: 35661172-35661172 on Build GRCh38
rs768700936 Chr.9: 35661116-35661116 on Build GRCh38
rs752108004 Chr.9: 35660984-35660984 on Build GRCh38
rs773360401 Chr.9: 35661126-35661126 on Build GRCh38
rs770854256 Chr.9: 35661175-35661176 on Build GRCh38
rs745464083 Chr.9: 35661092-35661092 on Build GRCh38
rs372621134 Chr.9: 35661094-35661094 on Build GRCh38
rs371120243 Chr.9: 35661184-35661184 on Build GRCh38
rs41298216 Chr.9: 35661147-35661147 on Build GRCh38
rs758064807 Chr.9: 35661162-35661162 on Build GRCh38
rs767970537 Chr.9: 35661072-35661072 on Build GRCh38
rs199991115 Chr.9: 35661058-35661058 on Build GRCh38
rs768717463 Chr.9: 35661057-35661057 on Build GRCh38
rs746812364 Chr.9: 35661115-35661115 on Build GRCh38
rs766603469 Chr.9: 35661135-35661135 on Build GRCh38
rs771140542 Chr.9: 35661195-35661195 on Build GRCh38

View Full Product Details