Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 30952073-30952342 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPCR1
Entrez Gene ID
135656
Gene Name
diffuse panbronchiolitis critical region 1
Gene Aliases
PBLT
UniGene
Hs.631993
Gene Chromosome Location
Chr.6: 30941000-30954221 on Build GRCh38
Transcripts
NM_080870.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3861613 c.257C>A p.P86H Chr.6: 30952195-30952195 on Build GRCh38
7365343 c.209C>A p.S70Y Chr.6: 30952147-30952147 on Build GRCh38
3702972 c.182C>T p.T61I Chr.6: 30952120-30952120 on Build GRCh38
7365342 c.1055C>A p.S352Y Chr.6: 30952147-30952147 on Build GRCh38
7365329 c.142G>C p.E48Q Chr.6: 30952080-30952080 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749215662 Chr.6: 30952234-30952234 on Build GRCh38
rs771778110 Chr.6: 30952256-30952256 on Build GRCh38
rs141271962 Chr.6: 30952177-30952177 on Build GRCh38
rs975361900 Chr.6: 30952109-30952109 on Build GRCh38
rs193206474 Chr.6: 30952250-30952250 on Build GRCh38
rs1002090854 Chr.6: 30952273-30952273 on Build GRCh38
rs775298792 Chr.6: 30952116-30952116 on Build GRCh38
rs879147245 Chr.6: 30952161-30952161 on Build GRCh38
rs55710655 Chr.6: 30952270-30952270 on Build GRCh38
rs765764467 Chr.6: 30952305-30952305 on Build GRCh38
rs200921598 Chr.6: 30952231-30952231 on Build GRCh38
rs79792575 Chr.6: 30952309-30952309 on Build GRCh38
rs767132860 Chr.6: 30952142-30952142 on Build GRCh38
rs750843443 Chr.6: 30952153-30952153 on Build GRCh38
rs11970154 Chr.6: 30952101-30952101 on Build GRCh38
rs571239037 Chr.6: 30952238-30952238 on Build GRCh38
rs766673062 Chr.6: 30952271-30952271 on Build GRCh38
rs139901951 Chr.6: 30952134-30952134 on Build GRCh38
rs1033534027 Chr.6: 30952278-30952278 on Build GRCh38
rs759999003 Chr.6: 30952289-30952289 on Build GRCh38
rs756466579 Chr.6: 30952164-30952164 on Build GRCh38
rs754309761 Chr.6: 30952176-30952176 on Build GRCh38
rs934120997 Chr.6: 30952133-30952133 on Build GRCh38
rs748218735 Chr.6: 30952200-30952200 on Build GRCh38
rs769922906 Chr.6: 30952107-30952107 on Build GRCh38
rs745648072 Chr.6: 30952102-30952102 on Build GRCh38
rs766696153 Chr.6: 30952167-30952167 on Build GRCh38
rs760259062 Chr.6: 30952266-30952266 on Build GRCh38
rs747165922 Chr.6: 30952218-30952218 on Build GRCh38
rs776864263 Chr.6: 30952145-30952145 on Build GRCh38
rs763535639 Chr.6: 30952123-30952123 on Build GRCh38
rs769489613 Chr.6: 30952230-30952230 on Build GRCh38
rs778887996 Chr.6: 30952193-30952193 on Build GRCh38
rs74442959 Chr.6: 30952098-30952098 on Build GRCh38
rs762892209 Chr.6: 30952117-30952117 on Build GRCh38
rs371020510 Chr.6: 30952128-30952128 on Build GRCh38
rs750822087 Chr.6: 30952195-30952196 on Build GRCh38
rs149982150 Chr.6: 30952310-30952310 on Build GRCh38
rs754188367 Chr.6: 30952272-30952272 on Build GRCh38
rs779602439 Chr.6: 30952132-30952132 on Build GRCh38
rs201694765 Chr.6: 30952206-30952206 on Build GRCh38
rs773780783 Chr.6: 30952244-30952244 on Build GRCh38
rs772856191 Chr.6: 30952260-30952260 on Build GRCh38
rs150846286 Chr.6: 30952215-30952215 on Build GRCh38

View Full Product Details