Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 20555282-20555525 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSM2B
Entrez Gene ID
348158
Gene Name
acyl-CoA synthetase medium-chain family member 2B
Gene Aliases
ACSM2, HXMA, HYST1046
UniGene
Hs.298252
Gene Chromosome Location
Chr.16: 20536261-20576379 on Build GRCh38
Transcripts
NM_001105069.1 NM_182617.3 XM_005255304.3 XM_011545824.2 XM_017023205.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7288040 c.500T>C p.V167A Chr.16: 20555365-20555365 on Build GRCh38
4420036 c.404C>A p.P135H Chr.16: 20555461-20555461 on Build GRCh38
6592688 c.533G>T p.R178I Chr.16: 20555332-20555332 on Build GRCh38
4897873 c.524C>T p.P175L Chr.16: 20555341-20555341 on Build GRCh38
4256924 c.420G>A p.M140I Chr.16: 20555445-20555445 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368213752 Chr.16: 20555485-20555485 on Build GRCh38
rs749463090 Chr.16: 20555354-20555354 on Build GRCh38
rs764879533 Chr.16: 20555483-20555483 on Build GRCh38
rs771075330 Chr.16: 20555355-20555355 on Build GRCh38
rs751510953 Chr.16: 20555423-20555423 on Build GRCh38
rs372297379 Chr.16: 20555486-20555486 on Build GRCh38
rs758361163 Chr.16: 20555326-20555326 on Build GRCh38
rs769904929 Chr.16: 20555342-20555342 on Build GRCh38
rs778780934 Chr.16: 20555322-20555322 on Build GRCh38
rs895136438 Chr.16: 20555382-20555382 on Build GRCh38
rs746992275 Chr.16: 20555330-20555330 on Build GRCh38
rs779079829 Chr.16: 20555452-20555452 on Build GRCh38
rs145882700 Chr.16: 20555439-20555439 on Build GRCh38
rs750256258 Chr.16: 20555325-20555325 on Build GRCh38
rs147789042 Chr.16: 20555464-20555464 on Build GRCh38
rs776156683 Chr.16: 20555479-20555479 on Build GRCh38
rs780053622 Chr.16: 20555414-20555414 on Build GRCh38
rs769054096 Chr.16: 20555471-20555471 on Build GRCh38
rs563386991 Chr.16: 20555492-20555492 on Build GRCh38
rs774653362 Chr.16: 20555359-20555359 on Build GRCh38
rs772469982 Chr.16: 20555385-20555385 on Build GRCh38
rs759928318 Chr.16: 20555373-20555373 on Build GRCh38
rs746141038 Chr.16: 20555455-20555455 on Build GRCh38
rs775949953 Chr.16: 20555392-20555392 on Build GRCh38
rs1055160229 Chr.16: 20555371-20555371 on Build GRCh38
rs753511934 Chr.16: 20555305-20555305 on Build GRCh38
rs559620196 Chr.16: 20555329-20555329 on Build GRCh38
rs761141010 Chr.16: 20555396-20555396 on Build GRCh38
rs753290502 Chr.16: 20555405-20555405 on Build GRCh38
rs754877764 Chr.16: 20555427-20555427 on Build GRCh38
rs756925225 Chr.16: 20555308-20555308 on Build GRCh38
rs574648610 Chr.16: 20555406-20555406 on Build GRCh38
rs750203074 Chr.16: 20555413-20555413 on Build GRCh38
rs111238231 Chr.16: 20555301-20555301 on Build GRCh38
rs771105536 Chr.16: 20555450-20555450 on Build GRCh38
rs761259571 Chr.16: 20555481-20555481 on Build GRCh38
rs751327335 Chr.16: 20555496-20555496 on Build GRCh38
rs777783548 Chr.16: 20555344-20555344 on Build GRCh38
rs777853409 Chr.16: 20555431-20555431 on Build GRCh38
rs763653546 Chr.16: 20555401-20555401 on Build GRCh38
rs772409041 Chr.16: 20555462-20555462 on Build GRCh38
rs374394716 Chr.16: 20555469-20555469 on Build GRCh38
rs1043723129 Chr.16: 20555438-20555438 on Build GRCh38

View Full Product Details