Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 45357229-45357472 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANO6
Entrez Gene ID
196527
Gene Name
anoctamin 6
Gene Aliases
BDPLT7, SCTS, TMEM16F
UniGene
Hs.505339
Gene Chromosome Location
Chr.12: 45215987-45440404 on Build GRCh38
Transcripts
NM_001025356.2 NM_001142678.1 NM_001142679.1 NM_001204803.1 XM_005268706.4 XM_005268707.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6587587 c.954T>G p.A318A Chr.12: 45357380-45357380 on Build GRCh38
6900305 c.877C>T p.L293L Chr.12: 45357357-45357357 on Build GRCh38
5619423 c.936C>T p.A312A Chr.12: 45357362-45357362 on Build GRCh38
7252512 c.996G>C p.W332C Chr.12: 45357422-45357422 on Build GRCh38
6601120 c.895T>G p.F299V Chr.12: 45357321-45357321 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745758616 Chr.12: 45357321-45357322 on Build GRCh38
rs767822179 Chr.12: 45357430-45357430 on Build GRCh38
rs940272748 Chr.12: 45357345-45357345 on Build GRCh38
rs745362044 Chr.12: 45357383-45357383 on Build GRCh38
rs774722327 Chr.12: 45357409-45357409 on Build GRCh38
rs917945450 Chr.12: 45357343-45357343 on Build GRCh38
rs770243770 Chr.12: 45357387-45357387 on Build GRCh38
rs901533782 Chr.12: 45357438-45357438 on Build GRCh38
rs763252734 Chr.12: 45357397-45357397 on Build GRCh38
rs776030027 Chr.12: 45357391-45357391 on Build GRCh38
rs767770827 Chr.12: 45357322-45357322 on Build GRCh38
rs199670117 Chr.12: 45357382-45357382 on Build GRCh38
rs763345289 Chr.12: 45357312-45357312 on Build GRCh38
rs138627790 Chr.12: 45357429-45357429 on Build GRCh38
rs747953092 Chr.12: 45357261-45357261 on Build GRCh38
rs779937111 Chr.12: 45357441-45357441 on Build GRCh38
rs750516037 Chr.12: 45357324-45357324 on Build GRCh38
rs751784217 Chr.12: 45357320-45357320 on Build GRCh38
rs888870828 Chr.12: 45357402-45357402 on Build GRCh38
rs146500373 Chr.12: 45357330-45357330 on Build GRCh38
rs771662255 Chr.12: 45357266-45357266 on Build GRCh38
rs778878497 Chr.12: 45357334-45357334 on Build GRCh38
rs756766801 Chr.12: 45357275-45357276 on Build GRCh38
rs145896287 Chr.12: 45357362-45357362 on Build GRCh38
rs746596449 Chr.12: 45357375-45357375 on Build GRCh38
rs750504458 Chr.12: 45357437-45357437 on Build GRCh38
rs778586886 Chr.12: 45357279-45357282 on Build GRCh38
rs866240802 Chr.12: 45357338-45357339 on Build GRCh38
rs760031599 Chr.12: 45357297-45357297 on Build GRCh38
rs569930058 Chr.12: 45357319-45357319 on Build GRCh38
rs772807445 Chr.12: 45357269-45357269 on Build GRCh38
rs763615507 Chr.12: 45357315-45357315 on Build GRCh38
rs770568121 Chr.12: 45357378-45357378 on Build GRCh38
rs926719236 Chr.12: 45357268-45357268 on Build GRCh38
rs772139610 Chr.12: 45357339-45357340 on Build GRCh38
rs145510962 Chr.12: 45357298-45357298 on Build GRCh38
rs955328263 Chr.12: 45357354-45357354 on Build GRCh38
rs747922978 Chr.12: 45357358-45357358 on Build GRCh38
rs920066485 Chr.12: 45357372-45357372 on Build GRCh38
rs371009341 Chr.12: 45357281-45357281 on Build GRCh38
rs775771097 Chr.12: 45357308-45357308 on Build GRCh38
rs764499243 Chr.12: 45357406-45357406 on Build GRCh38
rs777413788 Chr.12: 45357363-45357363 on Build GRCh38
rs777125229 Chr.12: 45357287-45357287 on Build GRCh38
rs778630561 Chr.12: 45357259-45357259 on Build GRCh38

View Full Product Details