Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 10221630-10221889 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABARAPL1
Entrez Gene ID
23710
Gene Name
GABA type A receptor associated protein like 1
Gene Aliases
APG8-LIKE, APG8L, ATG8, ATG8B, ATG8L, GEC1
UniGene
Hs.524250
Gene Chromosome Location
Chr.12: 10212841-10223128 on Build GRCh38
Transcripts
NM_031412.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4038119 c.289-1G>A p.? Chr.12: 10221786-10221786 on Build GRCh38
6562420 c.301_303delGAA p.E101delE Chr.12: 10221799-10221801 on Build GRCh38
4038120 c.313C>A p.L105M Chr.12: 10221811-10221811 on Build GRCh38
5630483 c.354A>G p.*118W Chr.12: 10221852-10221852 on Build GRCh38
6348273 c.329G>T p.S110I Chr.12: 10221827-10221827 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758603696 Chr.12: 10221758-10221758 on Build GRCh38
rs770747565 Chr.12: 10221851-10221851 on Build GRCh38
rs56206562 Chr.12: 10221836-10221836 on Build GRCh38
rs776545848 Chr.12: 10221854-10221854 on Build GRCh38
rs566450817 Chr.12: 10221679-10221679 on Build GRCh38
rs764240653 Chr.12: 10221782-10221782 on Build GRCh38
rs573249146 Chr.12: 10221759-10221759 on Build GRCh38
rs922180220 Chr.12: 10221780-10221780 on Build GRCh38
rs756492894 Chr.12: 10221817-10221817 on Build GRCh38
rs78452572 Chr.12: 10221674-10221674 on Build GRCh38
rs780385742 Chr.12: 10221819-10221819 on Build GRCh38
rs780977650 Chr.12: 10221806-10221806 on Build GRCh38
rs1044210340 Chr.12: 10221753-10221753 on Build GRCh38
rs748739155 Chr.12: 10221841-10221841 on Build GRCh38
rs749803834 Chr.12: 10221820-10221820 on Build GRCh38
rs746657006 Chr.12: 10221850-10221850 on Build GRCh38
rs146121739 Chr.12: 10221843-10221843 on Build GRCh38
rs963496251 Chr.12: 10221728-10221728 on Build GRCh38
rs751714337 Chr.12: 10221791-10221791 on Build GRCh38
rs374836231 Chr.12: 10221857-10221857 on Build GRCh38
rs777401178 Chr.12: 10221849-10221849 on Build GRCh38
rs139914471 Chr.12: 10221870-10221870 on Build GRCh38
rs757539231 Chr.12: 10221795-10221795 on Build GRCh38
rs1015025496 Chr.12: 10221683-10221683 on Build GRCh38
rs750831411 Chr.12: 10221807-10221807 on Build GRCh38
rs953419407 Chr.12: 10221781-10221781 on Build GRCh38

View Full Product Details