Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 219280501-219280774 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJB2
Entrez Gene ID
3300
Gene Name
DnaJ heat shock protein family (Hsp40) member B2
Gene Aliases
CMT2T, DSMA5, HSJ-1, HSJ1, HSPF3
UniGene
Hs.77768
Gene Chromosome Location
Chr.2: 219279318-219286900 on Build GRCh38
Transcripts
NM_001039550.1 NM_006736.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4091652 c.73C>T p.R25C Chr.2: 219280585-219280585 on Build GRCh38
7144655 c.150C>T p.A50A Chr.2: 219280662-219280662 on Build GRCh38
6424941 c.97G>A p.D33N Chr.2: 219280609-219280609 on Build GRCh38
1129317 c.74G>A p.R25H Chr.2: 219280586-219280586 on Build GRCh38
6424556 c.95C>A p.P32Q Chr.2: 219280607-219280607 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761058767 Chr.2: 219280682-219280682 on Build GRCh38
rs755728990 Chr.2: 219280618-219280618 on Build GRCh38
rs774792381 Chr.2: 219280543-219280543 on Build GRCh38
rs780839621 Chr.2: 219280583-219280583 on Build GRCh38
rs776887529 Chr.2: 219280696-219280696 on Build GRCh38
rs774489758 Chr.2: 219280704-219280704 on Build GRCh38
rs773451478 Chr.2: 219280680-219280680 on Build GRCh38
rs370819068 Chr.2: 219280582-219280582 on Build GRCh38
rs562669797 Chr.2: 219280689-219280689 on Build GRCh38
rs768122577 Chr.2: 219280661-219280661 on Build GRCh38
rs200212471 Chr.2: 219280713-219280713 on Build GRCh38
rs753872606 Chr.2: 219280574-219280574 on Build GRCh38
rs767423929 Chr.2: 219280734-219280734 on Build GRCh38
rs748638654 Chr.2: 219280668-219280668 on Build GRCh38
rs2276638 Chr.2: 219280564-219280564 on Build GRCh38
rs936213383 Chr.2: 219280630-219280630 on Build GRCh38
rs748820330 Chr.2: 219280636-219280636 on Build GRCh38
rs770139282 Chr.2: 219280550-219280551 on Build GRCh38
rs912231859 Chr.2: 219280725-219280725 on Build GRCh38
rs117458162 Chr.2: 219280560-219280560 on Build GRCh38
rs779463890 Chr.2: 219280662-219280662 on Build GRCh38
rs375470108 Chr.2: 219280700-219280700 on Build GRCh38
rs772494708 Chr.2: 219280697-219280697 on Build GRCh38
rs753121404 Chr.2: 219280735-219280735 on Build GRCh38
rs1033804318 Chr.2: 219280557-219280557 on Build GRCh38
rs539825713 Chr.2: 219280554-219280554 on Build GRCh38
rs759733377 Chr.2: 219280699-219280699 on Build GRCh38
rs375566786 Chr.2: 219280547-219280547 on Build GRCh38
rs748320610 Chr.2: 219280554-219280556 on Build GRCh38
rs781334383 Chr.2: 219280550-219280553 on Build GRCh38
rs548806487 Chr.2: 219280622-219280622 on Build GRCh38
rs1021860602 Chr.2: 219280624-219280624 on Build GRCh38
rs762203771 Chr.2: 219280546-219280546 on Build GRCh38
rs772508907 Chr.2: 219280670-219280670 on Build GRCh38
rs750175376 Chr.2: 219280585-219280585 on Build GRCh38
rs751515788 Chr.2: 219280575-219280575 on Build GRCh38
rs967333105 Chr.2: 219280627-219280630 on Build GRCh38

View Full Product Details