Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 62615654-62615889 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B3GAT3
Entrez Gene ID
26229
Gene Name
beta-1,3-glucuronyltransferase 3
Gene Aliases
GLCATI, JDSCD, glcUAT-I
UniGene
Hs.502759
Gene Chromosome Location
Chr.11: 62615296-62622175 on Build GRCh38
Transcripts
NM_001288721.1 NM_001288722.1 NM_001288723.1 NM_012200.3 NR_109991.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3451038 c.926C>T p.T309I Chr.11: 62615783-62615783 on Build GRCh38
5985325 c.920G>C p.W307S Chr.11: 62615789-62615789 on Build GRCh38
2039095 c.970C>T p.R324W Chr.11: 62615739-62615739 on Build GRCh38
5572169 c.919T>C p.W307R Chr.11: 62615790-62615790 on Build GRCh38
429452 c.928C>T p.R310W Chr.11: 62615781-62615781 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs764917865 Chr.11: 62615697-62615697 on Build GRCh38
rs775090938 Chr.11: 62615707-62615707 on Build GRCh38
rs767784957 Chr.11: 62615694-62615696 on Build GRCh38
rs201436267 Chr.11: 62615683-62615683 on Build GRCh38
rs749435133 Chr.11: 62615751-62615751 on Build GRCh38
rs368250687 Chr.11: 62615859-62615859 on Build GRCh38
rs527748687 Chr.11: 62615853-62615853 on Build GRCh38
rs904110417 Chr.11: 62615829-62615829 on Build GRCh38
rs147883248 Chr.11: 62615779-62615779 on Build GRCh38
rs1042651254 Chr.11: 62615808-62615808 on Build GRCh38
rs200511074 Chr.11: 62615730-62615730 on Build GRCh38
rs774081276 Chr.11: 62615839-62615839 on Build GRCh38
rs748537810 Chr.11: 62615801-62615801 on Build GRCh38
rs376610156 Chr.11: 62615696-62615696 on Build GRCh38
rs750535749 Chr.11: 62615717-62615717 on Build GRCh38
rs768503650 Chr.11: 62615838-62615838 on Build GRCh38
rs369154163 Chr.11: 62615738-62615738 on Build GRCh38
rs779076359 Chr.11: 62615780-62615780 on Build GRCh38
rs143252096 Chr.11: 62615860-62615860 on Build GRCh38
rs770428920 Chr.11: 62615684-62615684 on Build GRCh38
rs199603266 Chr.11: 62615809-62615809 on Build GRCh38
rs531719501 Chr.11: 62615727-62615727 on Build GRCh38
rs775230481 Chr.11: 62615834-62615834 on Build GRCh38
rs945030545 Chr.11: 62615795-62615795 on Build GRCh38
rs11554366 Chr.11: 62615686-62615686 on Build GRCh38
rs778227646 Chr.11: 62615806-62615806 on Build GRCh38
rs1017642684 Chr.11: 62615731-62615731 on Build GRCh38
rs762837360 Chr.11: 62615709-62615709 on Build GRCh38
rs762196321 Chr.11: 62615693-62615694 on Build GRCh38
rs926444789 Chr.11: 62615677-62615677 on Build GRCh38
rs201387537 Chr.11: 62615802-62615802 on Build GRCh38
rs762455251 Chr.11: 62615836-62615836 on Build GRCh38
rs769251506 Chr.11: 62615826-62615826 on Build GRCh38
rs745422107 Chr.11: 62615817-62615817 on Build GRCh38
rs569989507 Chr.11: 62615729-62615729 on Build GRCh38
rs868665260 Chr.11: 62615781-62615781 on Build GRCh38
rs763747195 Chr.11: 62615711-62615711 on Build GRCh38
rs774847138 Chr.11: 62615691-62615691 on Build GRCh38
rs780467028 Chr.11: 62615739-62615739 on Build GRCh38

View Full Product Details