Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 224952606-224952875 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAH14
Entrez Gene ID
127602
Gene Name
dynein axonemal heavy chain 14
Gene Aliases
C1orf67, Dnahc14, HL-18, HL18
UniGene
Hs.133977
Gene Chromosome Location
Chr.1: 224928422-225399294 on Build GRCh38
Transcripts
NM_001145154.1 NM_001373.1 NM_144989.2 XM_006711735.3 XM_011544057.2 XM_011544058.2 XM_011544059.2 XM_011544061.2 XM_011544062.2 XM_011544063.2 XM_011544064.2 XM_011544065.2 XM_011544066.2 XM_011544067.2 XM_011544069.2 XM_011544070.2 XM_011544071.2 XM_011544072.2 XM_017000295.1 XM_017000296.1 XM_017000297.1 XM_017000298.1 XM_017000300.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4143387 c.63C>A p.T21T Chr.1: 224952765-224952765 on Build GRCh38
3484195 c.571C>T p.Q191* Chr.1: 224952742-224952742 on Build GRCh38
6435765 c.45G>T p.E15D Chr.1: 224952747-224952747 on Build GRCh38
1960921 c.537G>A p.E179E Chr.1: 224952708-224952708 on Build GRCh38
4143390 c.64A>T p.K22* Chr.1: 224952766-224952766 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761635864 Chr.1: 224952678-224952678 on Build GRCh38
rs866628060 Chr.1: 224952766-224952766 on Build GRCh38
rs914841683 Chr.1: 224952806-224952806 on Build GRCh38
rs1013185875 Chr.1: 224952685-224952685 on Build GRCh38
rs768612537 Chr.1: 224952669-224952669 on Build GRCh38
rs756479434 Chr.1: 224952756-224952756 on Build GRCh38
rs367747039 Chr.1: 224952791-224952791 on Build GRCh38
rs781743868 Chr.1: 224952752-224952752 on Build GRCh38
rs750980879 Chr.1: 224952753-224952753 on Build GRCh38
rs368164810 Chr.1: 224952653-224952653 on Build GRCh38
rs905416661 Chr.1: 224952749-224952749 on Build GRCh38
rs780365095 Chr.1: 224952663-224952663 on Build GRCh38
rs989797893 Chr.1: 224952803-224952803 on Build GRCh38
rs569798175 Chr.1: 224952699-224952699 on Build GRCh38
rs201634729 Chr.1: 224952769-224952769 on Build GRCh38
rs764641745 Chr.1: 224952723-224952723 on Build GRCh38
rs753425198 Chr.1: 224952710-224952710 on Build GRCh38
rs1039632559 Chr.1: 224952823-224952823 on Build GRCh38
rs757875157 Chr.1: 224952745-224952745 on Build GRCh38
rs924333528 Chr.1: 224952719-224952719 on Build GRCh38
rs570666767 Chr.1: 224952764-224952764 on Build GRCh38
rs778980313 Chr.1: 224952694-224952695 on Build GRCh38
rs192193099 Chr.1: 224952711-224952711 on Build GRCh38
rs748219208 Chr.1: 224952787-224952787 on Build GRCh38
rs772087936 Chr.1: 224952684-224952684 on Build GRCh38
rs752289528 Chr.1: 224952739-224952739 on Build GRCh38
rs976738221 Chr.1: 224952833-224952833 on Build GRCh38
rs776832738 Chr.1: 224952801-224952801 on Build GRCh38
rs61745064 Chr.1: 224952757-224952757 on Build GRCh38
rs553800146 Chr.1: 224952810-224952810 on Build GRCh38
rs538639998 Chr.1: 224952765-224952765 on Build GRCh38
rs773080212 Chr.1: 224952799-224952799 on Build GRCh38
rs778829039 Chr.1: 224952771-224952771 on Build GRCh38
rs766179429 Chr.1: 224952708-224952708 on Build GRCh38
rs546294120 Chr.1: 224952837-224952837 on Build GRCh38
rs774545761 Chr.1: 224952676-224952676 on Build GRCh38
rs764949097 Chr.1: 224952815-224952815 on Build GRCh38
rs760412751 Chr.1: 224952701-224952701 on Build GRCh38
rs746158724 Chr.1: 224952793-224952793 on Build GRCh38
rs879071205 Chr.1: 224952637-224952637 on Build GRCh38
rs754722708 Chr.1: 224952798-224952800 on Build GRCh38
rs757394805 Chr.1: 224952687-224952689 on Build GRCh38
rs749528863 Chr.1: 224952667-224952667 on Build GRCh38

View Full Product Details