Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 49323568-49323833 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C10orf71
Entrez Gene ID
118461
Gene Name
chromosome 10 open reading frame 71
Gene Aliases
-
UniGene
Hs.585480
Gene Chromosome Location
Chr.10: 49297009-49330145 on Build GRCh38
Transcripts
NM_001135196.1 XM_005269476.4 XM_005269477.3 XM_005269478.4 XM_017015619.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3867333 c.1117C>T p.Q373* Chr.10: 49323662-49323662 on Build GRCh38
4650941 c.1105C>A p.Q369K Chr.10: 49323650-49323650 on Build GRCh38
5806579 c.1221A>T p.R407S Chr.10: 49323766-49323766 on Build GRCh38
3438341 c.1222G>A p.G408S Chr.10: 49323767-49323767 on Build GRCh38
4901368 c.1265C>T p.S422L Chr.10: 49323810-49323810 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1027893590 Chr.10: 49323679-49323679 on Build GRCh38
rs755271393 Chr.10: 49323795-49323795 on Build GRCh38
rs372639784 Chr.10: 49323676-49323676 on Build GRCh38
rs757907021 Chr.10: 49323775-49323775 on Build GRCh38
rs985271678 Chr.10: 49323633-49323633 on Build GRCh38
rs751283820 Chr.10: 49323794-49323794 on Build GRCh38
rs569250483 Chr.10: 49323733-49323733 on Build GRCh38
rs763303352 Chr.10: 49323640-49323640 on Build GRCh38
rs773543042 Chr.10: 49323626-49323626 on Build GRCh38
rs768662308 Chr.10: 49323803-49323803 on Build GRCh38
rs757999079 Chr.10: 49323656-49323656 on Build GRCh38
rs1050520740 Chr.10: 49323762-49323763 on Build GRCh38
rs556783319 Chr.10: 49323643-49323643 on Build GRCh38
rs748396302 Chr.10: 49323707-49323707 on Build GRCh38
rs780358490 Chr.10: 49323590-49323590 on Build GRCh38
rs777198549 Chr.10: 49323597-49323597 on Build GRCh38
rs1056243152 Chr.10: 49323613-49323613 on Build GRCh38
rs182247538 Chr.10: 49323671-49323671 on Build GRCh38
rs766015437 Chr.10: 49323777-49323777 on Build GRCh38
rs748172824 Chr.10: 49323609-49323609 on Build GRCh38
rs758382540 Chr.10: 49323652-49323652 on Build GRCh38
rs765182836 Chr.10: 49323769-49323769 on Build GRCh38
rs781647570 Chr.10: 49323796-49323796 on Build GRCh38
rs749875321 Chr.10: 49323771-49323771 on Build GRCh38
rs749064221 Chr.10: 49323677-49323677 on Build GRCh38
rs770052650 Chr.10: 49323614-49323614 on Build GRCh38
rs1037140257 Chr.10: 49323646-49323646 on Build GRCh38
rs748734187 Chr.10: 49323682-49323682 on Build GRCh38
rs749969036 Chr.10: 49323654-49323654 on Build GRCh38
rs756638573 Chr.10: 49323802-49323802 on Build GRCh38
rs763937884 Chr.10: 49323644-49323644 on Build GRCh38
rs763998364 Chr.10: 49323760-49323760 on Build GRCh38
rs776402238 Chr.10: 49323761-49323761 on Build GRCh38
rs773383651 Chr.10: 49323688-49323688 on Build GRCh38
rs774705149 Chr.10: 49323744-49323744 on Build GRCh38
rs185740154 Chr.10: 49323680-49323680 on Build GRCh38
rs748489790 Chr.10: 49323798-49323798 on Build GRCh38
rs866306696 Chr.10: 49323744-49323744 on Build GRCh38
rs896197542 Chr.10: 49323641-49323641 on Build GRCh38
rs769959614 Chr.10: 49323686-49323686 on Build GRCh38
rs771213703 Chr.10: 49323738-49323738 on Build GRCh38
rs375386226 Chr.10: 49323804-49323804 on Build GRCh38
rs201571697 Chr.10: 49323642-49323642 on Build GRCh38
rs778984450 Chr.10: 49323807-49323807 on Build GRCh38
rs761772075 Chr.10: 49323765-49323765 on Build GRCh38
rs367577636 Chr.10: 49323648-49323648 on Build GRCh38
rs373739260 Chr.10: 49323595-49323595 on Build GRCh38
rs760379065 Chr.10: 49323757-49323757 on Build GRCh38

View Full Product Details