Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 23266324-23266559 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPNMB
Entrez Gene ID
10457
Gene Name
glycoprotein nmb
Gene Aliases
HGFIN, NMB
UniGene
Hs.190495
Gene Chromosome Location
Chr.7: 23246206-23275110 on Build GRCh38
Transcripts
NM_001005340.1 NM_002510.2 XM_005249578.2 XM_017011676.1 XM_017011677.1 XM_017011678.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1635249 c.1052C>T p.P351L Chr.7: 23266514-23266514 on Build GRCh38
7303089 c.1019-5C>T p.? Chr.7: 23266476-23266476 on Build GRCh38
6109701 c.1088G>A p.R363K Chr.7: 23266550-23266550 on Build GRCh38
3768325 c.1081C>T p.L361L Chr.7: 23266543-23266543 on Build GRCh38
3880270 c.1056A>G p.G352G Chr.7: 23266518-23266518 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777617425 Chr.7: 23266533-23266533 on Build GRCh38
rs762304625 Chr.7: 23266471-23266471 on Build GRCh38
rs75801644 Chr.7: 23266522-23266522 on Build GRCh38
rs774552337 Chr.7: 23266462-23266462 on Build GRCh38
rs768088459 Chr.7: 23266477-23266477 on Build GRCh38
rs886894610 Chr.7: 23266518-23266518 on Build GRCh38
rs201506520 Chr.7: 23266458-23266458 on Build GRCh38
rs927253214 Chr.7: 23266445-23266445 on Build GRCh38
rs866339862 Chr.7: 23266513-23266513 on Build GRCh38
rs201810730 Chr.7: 23266505-23266505 on Build GRCh38
rs76375148 Chr.7: 23266358-23266358 on Build GRCh38
rs766366366 Chr.7: 23266500-23266500 on Build GRCh38
rs1010102645 Chr.7: 23266381-23266381 on Build GRCh38
rs752866089 Chr.7: 23266489-23266490 on Build GRCh38
rs770915595 Chr.7: 23266536-23266536 on Build GRCh38
rs1032893132 Chr.7: 23266369-23266369 on Build GRCh38
rs779226838 Chr.7: 23266507-23266507 on Build GRCh38
rs156429 Chr.7: 23266401-23266401 on Build GRCh38
rs759101024 Chr.7: 23266430-23266430 on Build GRCh38
rs373107581 Chr.7: 23266419-23266419 on Build GRCh38
rs184014301 Chr.7: 23266537-23266537 on Build GRCh38
rs539479734 Chr.7: 23266426-23266426 on Build GRCh38
rs769538102 Chr.7: 23266442-23266442 on Build GRCh38
rs943964978 Chr.7: 23266365-23266365 on Build GRCh38
rs552546907 Chr.7: 23266402-23266402 on Build GRCh38
rs191551544 Chr.7: 23266530-23266530 on Build GRCh38
rs957427076 Chr.7: 23266377-23266377 on Build GRCh38
rs753996624 Chr.7: 23266502-23266502 on Build GRCh38
rs746942058 Chr.7: 23266534-23266534 on Build GRCh38
rs760771359 Chr.7: 23266486-23266486 on Build GRCh38
rs1020773181 Chr.7: 23266382-23266382 on Build GRCh38
rs750981123 Chr.7: 23266480-23266480 on Build GRCh38

View Full Product Details