Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 168765647-168765886 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CERS6
Entrez Gene ID
253782
Gene Name
ceramide synthase 6
Gene Aliases
CERS5, LASS6
UniGene
Hs.506829
Gene Chromosome Location
Chr.2: 168456249-168775134 on Build GRCh38
Transcripts
NM_001256126.1 NM_203463.2 XM_005246440.4 XM_017003749.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1669384 c.934C>T p.Q312* Chr.2: 168765680-168765680 on Build GRCh38
1193392 c.972A>T p.K324N Chr.2: 168765718-168765718 on Build GRCh38
6873487 c.939G>A p.G313G Chr.2: 168765685-168765685 on Build GRCh38
1009073 c.998G>A p.G333D Chr.2: 168765744-168765744 on Build GRCh38
5993910 c.936A>C p.Q312H Chr.2: 168765682-168765682 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755716982 Chr.2: 168765787-168765787 on Build GRCh38
rs373773691 Chr.2: 168765763-168765763 on Build GRCh38
rs981193951 Chr.2: 168765830-168765830 on Build GRCh38
rs1029584108 Chr.2: 168765705-168765705 on Build GRCh38
rs919545392 Chr.2: 168765772-168765772 on Build GRCh38
rs374085466 Chr.2: 168765682-168765682 on Build GRCh38
rs955387395 Chr.2: 168765748-168765748 on Build GRCh38
rs367638082 Chr.2: 168765754-168765754 on Build GRCh38
rs770689483 Chr.2: 168765798-168765798 on Build GRCh38
rs948887154 Chr.2: 168765849-168765849 on Build GRCh38
rs761996415 Chr.2: 168765678-168765678 on Build GRCh38
rs776890454 Chr.2: 168765674-168765674 on Build GRCh38
rs765701588 Chr.2: 168765684-168765684 on Build GRCh38
rs750742526 Chr.2: 168765730-168765730 on Build GRCh38
rs72881726 Chr.2: 168765822-168765822 on Build GRCh38
rs763320618 Chr.2: 168765736-168765736 on Build GRCh38
rs918788029 Chr.2: 168765816-168765816 on Build GRCh38
rs114643607 Chr.2: 168765792-168765792 on Build GRCh38
rs963318529 Chr.2: 168765782-168765782 on Build GRCh38
rs186536229 Chr.2: 168765756-168765756 on Build GRCh38
rs902596266 Chr.2: 168765675-168765675 on Build GRCh38
rs780826771 Chr.2: 168765760-168765760 on Build GRCh38
rs766802170 Chr.2: 168765753-168765753 on Build GRCh38

View Full Product Details