Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 48187103-48187372 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRMPD2
Entrez Gene ID
143162
Gene Name
FERM and PDZ domain containing 2
Gene Aliases
PDZD5C, PDZK4, PDZK5C
UniGene
Hs.664786
Gene Chromosome Location
Chr.10: 48156559-48274898 on Build GRCh38
Transcripts
NM_001018071.3 NM_001318191.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3438183 c.2220G>A p.M740I Chr.10: 48187238-48187238 on Build GRCh38
4516675 c.2180_2181GG>AA p.R727Q Chr.10: 48187277-48187278 on Build GRCh38
2144361 c.2233C>T p.L745F Chr.10: 48187225-48187225 on Build GRCh38
3438184 c.2176G>T p.G726C Chr.10: 48187282-48187282 on Build GRCh38
1347942 c.2237_2238delCT p.S746fs*14 Chr.10: 48187220-48187221 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs912037588 Chr.10: 48187196-48187196 on Build GRCh38
rs774364272 Chr.10: 48187328-48187328 on Build GRCh38
rs530514845 Chr.10: 48187333-48187333 on Build GRCh38
rs376996416 Chr.10: 48187335-48187335 on Build GRCh38
rs761053648 Chr.10: 48187265-48187265 on Build GRCh38
rs993796626 Chr.10: 48187144-48187144 on Build GRCh38
rs756565234 Chr.10: 48187176-48187176 on Build GRCh38
rs770965844 Chr.10: 48187229-48187229 on Build GRCh38
rs371554767 Chr.10: 48187220-48187220 on Build GRCh38
rs751136822 Chr.10: 48187164-48187164 on Build GRCh38
rs115290078 Chr.10: 48187297-48187297 on Build GRCh38
rs780280878 Chr.10: 48187217-48187217 on Build GRCh38
rs773165813 Chr.10: 48187154-48187154 on Build GRCh38
rs148821357 Chr.10: 48187239-48187239 on Build GRCh38
rs1032966866 Chr.10: 48187227-48187227 on Build GRCh38
rs763761421 Chr.10: 48187258-48187259 on Build GRCh38
rs543871423 Chr.10: 48187136-48187136 on Build GRCh38
rs964593429 Chr.10: 48187180-48187180 on Build GRCh38
rs367587429 Chr.10: 48187254-48187254 on Build GRCh38
rs762460365 Chr.10: 48187160-48187160 on Build GRCh38
rs761649143 Chr.10: 48187147-48187147 on Build GRCh38
rs750421128 Chr.10: 48187289-48187289 on Build GRCh38
rs559703505 Chr.10: 48187274-48187274 on Build GRCh38
rs944950021 Chr.10: 48187200-48187200 on Build GRCh38
rs769581629 Chr.10: 48187221-48187221 on Build GRCh38
rs988073636 Chr.10: 48187256-48187256 on Build GRCh38
rs767562950 Chr.10: 48187162-48187162 on Build GRCh38
rs1053079016 Chr.10: 48187211-48187211 on Build GRCh38
rs749145772 Chr.10: 48187324-48187324 on Build GRCh38
rs368335151 Chr.10: 48187170-48187170 on Build GRCh38
rs765115126 Chr.10: 48187150-48187150 on Build GRCh38
rs759131460 Chr.10: 48187236-48187236 on Build GRCh38
rs778380389 Chr.10: 48187183-48187183 on Build GRCh38
rs375803706 Chr.10: 48187230-48187230 on Build GRCh38
rs180943552 Chr.10: 48187298-48187298 on Build GRCh38
rs575785799 Chr.10: 48187306-48187306 on Build GRCh38
rs114243389 Chr.10: 48187194-48187194 on Build GRCh38
rs142488456 Chr.10: 48187278-48187278 on Build GRCh38
rs778695323 Chr.10: 48187212-48187212 on Build GRCh38
rs186121284 Chr.10: 48187348-48187348 on Build GRCh38
rs778857395 Chr.10: 48187195-48187195 on Build GRCh38
rs562480241 Chr.10: 48187138-48187138 on Build GRCh38
rs770748495 Chr.10: 48187325-48187325 on Build GRCh38
rs188103401 Chr.10: 48187175-48187175 on Build GRCh38
rs758536691 Chr.10: 48187292-48187292 on Build GRCh38
rs541696678 Chr.10: 48187226-48187226 on Build GRCh38
rs772165206 Chr.10: 48187202-48187202 on Build GRCh38
rs777537708 Chr.10: 48187320-48187320 on Build GRCh38
rs746088401 Chr.10: 48187142-48187142 on Build GRCh38
rs758653639 Chr.10: 48187220-48187221 on Build GRCh38
rs372129458 Chr.10: 48187146-48187146 on Build GRCh38
rs773077942 Chr.10: 48187225-48187225 on Build GRCh38
rs754581378 Chr.10: 48187165-48187165 on Build GRCh38
rs375274290 Chr.10: 48187332-48187332 on Build GRCh38
rs115907611 Chr.10: 48187279-48187279 on Build GRCh38

View Full Product Details