Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 107387627-107387882 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFNA5
Entrez Gene ID
1946
Gene Name
ephrin A5
Gene Aliases
AF1, EFL5, EPLG7, GLC1M, LERK7, RAGS
UniGene
Hs.288741
Gene Chromosome Location
Chr.5: 107376889-107672364 on Build GRCh38
Transcripts
NM_001962.2 XM_006714565.2 XM_011543250.2 XM_011543251.2 XM_017009205.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4910380 c.475A>G p.R159G Chr.5: 107387715-107387715 on Build GRCh38
3607462 c.462C>G p.L154L Chr.5: 107387728-107387728 on Build GRCh38
6652076 c.448T>G p.S150A Chr.5: 107387742-107387742 on Build GRCh38
6652077 c.453T>C p.C151C Chr.5: 107387737-107387737 on Build GRCh38
6168579 c.474G>T p.V158V Chr.5: 107387716-107387716 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372771476 Chr.5: 107387798-107387798 on Build GRCh38
rs768385117 Chr.5: 107387665-107387665 on Build GRCh38
rs373016513 Chr.5: 107387819-107387819 on Build GRCh38
rs761420086 Chr.5: 107387667-107387667 on Build GRCh38
rs745771522 Chr.5: 107387721-107387721 on Build GRCh38
rs751584235 Chr.5: 107387737-107387737 on Build GRCh38
rs369964187 Chr.5: 107387664-107387664 on Build GRCh38
rs184031419 Chr.5: 107387843-107387843 on Build GRCh38
rs1008739951 Chr.5: 107387756-107387756 on Build GRCh38
rs527563363 Chr.5: 107387812-107387814 on Build GRCh38
rs781318381 Chr.5: 107387752-107387752 on Build GRCh38
rs757317885 Chr.5: 107387744-107387744 on Build GRCh38
rs772894239 Chr.5: 107387821-107387821 on Build GRCh38
rs775962970 Chr.5: 107387659-107387659 on Build GRCh38
rs375415062 Chr.5: 107387688-107387688 on Build GRCh38
rs372250540 Chr.5: 107387687-107387687 on Build GRCh38
rs1055248307 Chr.5: 107387770-107387770 on Build GRCh38
rs747697726 Chr.5: 107387785-107387788 on Build GRCh38
rs1018340953 Chr.5: 107387764-107387764 on Build GRCh38
rs545063107 Chr.5: 107387652-107387652 on Build GRCh38
rs369853743 Chr.5: 107387711-107387711 on Build GRCh38
rs771409651 Chr.5: 107387759-107387759 on Build GRCh38
rs772766764 Chr.5: 107387820-107387822 on Build GRCh38
rs746331255 Chr.5: 107387754-107387754 on Build GRCh38
rs1046246244 Chr.5: 107387842-107387842 on Build GRCh38
rs777205754 Chr.5: 107387791-107387791 on Build GRCh38
rs143858187 Chr.5: 107387647-107387647 on Build GRCh38
rs188151039 Chr.5: 107387844-107387844 on Build GRCh38
rs759461866 Chr.5: 107387830-107387830 on Build GRCh38
rs200515632 Chr.5: 107387666-107387666 on Build GRCh38
rs756694734 Chr.5: 107387775-107387775 on Build GRCh38
rs780240959 Chr.5: 107387776-107387776 on Build GRCh38
rs753693853 Chr.5: 107387699-107387699 on Build GRCh38
rs949265395 Chr.5: 107387793-107387793 on Build GRCh38
rs767192239 Chr.5: 107387676-107387676 on Build GRCh38
rs768134361 Chr.5: 107387785-107387785 on Build GRCh38
rs1042121425 Chr.5: 107387710-107387710 on Build GRCh38
rs764308705 Chr.5: 107387726-107387726 on Build GRCh38
rs766061649 Chr.5: 107387698-107387698 on Build GRCh38
rs750378964 Chr.5: 107387680-107387680 on Build GRCh38
rs778374511 Chr.5: 107387784-107387785 on Build GRCh38
rs760703715 Chr.5: 107387692-107387692 on Build GRCh38

View Full Product Details