Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 89863226-89863479 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASP8AP2
Entrez Gene ID
9994
Gene Name
caspase 8 associated protein 2
Gene Aliases
CED-4, FLASH, RIP25
UniGene
Hs.558218
Gene Chromosome Location
Chr.6: 89829900-89874436 on Build GRCh38
Transcripts
NM_001137667.1 NM_001137668.1 NM_012115.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7286203 c.1750G>A p.D584N Chr.6: 89863459-89863459 on Build GRCh38
5880401 c.1525A>G p.T509A Chr.6: 89863234-89863234 on Build GRCh38
484449 c.1685T>A p.L562* Chr.6: 89863394-89863394 on Build GRCh38
6243029 c.1721T>C p.V574A Chr.6: 89863430-89863430 on Build GRCh38
5622979 c.1591G>A p.E531K Chr.6: 89863300-89863300 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776525910 Chr.6: 89863363-89863365 on Build GRCh38
rs747322163 Chr.6: 89863264-89863264 on Build GRCh38
rs780772034 Chr.6: 89863336-89863336 on Build GRCh38
rs776783124 Chr.6: 89863285-89863285 on Build GRCh38
rs377395999 Chr.6: 89863377-89863377 on Build GRCh38
rs745644840 Chr.6: 89863354-89863354 on Build GRCh38
rs374581994 Chr.6: 89863327-89863327 on Build GRCh38
rs768692342 Chr.6: 89863274-89863274 on Build GRCh38
rs537195378 Chr.6: 89863307-89863307 on Build GRCh38
rs989709126 Chr.6: 89863320-89863320 on Build GRCh38
rs755849439 Chr.6: 89863364-89863364 on Build GRCh38
rs751029331 Chr.6: 89863360-89863360 on Build GRCh38
rs377095494 Chr.6: 89863449-89863449 on Build GRCh38
rs200323558 Chr.6: 89863259-89863259 on Build GRCh38
rs370445193 Chr.6: 89863425-89863425 on Build GRCh38
rs756757960 Chr.6: 89863338-89863338 on Build GRCh38
rs767369972 Chr.6: 89863301-89863301 on Build GRCh38
rs976754407 Chr.6: 89863310-89863310 on Build GRCh38
rs558220001 Chr.6: 89863318-89863318 on Build GRCh38
rs570370253 Chr.6: 89863373-89863373 on Build GRCh38
rs761748100 Chr.6: 89863293-89863293 on Build GRCh38
rs370348129 Chr.6: 89863387-89863387 on Build GRCh38
rs966358767 Chr.6: 89863306-89863306 on Build GRCh38
rs781228496 Chr.6: 89863436-89863436 on Build GRCh38
rs778543947 Chr.6: 89863347-89863347 on Build GRCh38
rs376777596 Chr.6: 89863315-89863315 on Build GRCh38
rs759443523 Chr.6: 89863298-89863298 on Build GRCh38
rs570121757 Chr.6: 89863446-89863446 on Build GRCh38
rs774308719 Chr.6: 89863297-89863297 on Build GRCh38
rs370049023 Chr.6: 89863321-89863321 on Build GRCh38
rs200220681 Chr.6: 89863427-89863427 on Build GRCh38

View Full Product Details