Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 135160282-135160543 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CT55
Entrez Gene ID
54967
Gene Name
cancer/testis antigen 55
Gene Aliases
BJHCC20A, CXorf48
UniGene
Hs.272804
Gene Chromosome Location
Chr.X: 135156536-135171827 on Build GRCh38
Transcripts
NM_001031705.2 NM_017863.2 XM_005262427.1

SNP Detail

SNP ID Chromosome Location
rs782283864 Chr.X: 135160429-135160429 on Build GRCh38
rs782221386 Chr.X: 135160459-135160459 on Build GRCh38
rs189681081 Chr.X: 135160385-135160385 on Build GRCh38
rs782745176 Chr.X: 135160393-135160393 on Build GRCh38
rs782750170 Chr.X: 135160325-135160325 on Build GRCh38
rs781857817 Chr.X: 135160318-135160318 on Build GRCh38
rs782748208 Chr.X: 135160307-135160307 on Build GRCh38
rs781995580 Chr.X: 135160491-135160491 on Build GRCh38
rs782365800 Chr.X: 135160473-135160473 on Build GRCh38
rs782402986 Chr.X: 135160521-135160521 on Build GRCh38
rs782443990 Chr.X: 135160445-135160445 on Build GRCh38
rs782512322 Chr.X: 135160423-135160423 on Build GRCh38
rs374625968 Chr.X: 135160426-135160426 on Build GRCh38
rs3813155 Chr.X: 135160400-135160400 on Build GRCh38
rs782266435 Chr.X: 135160501-135160501 on Build GRCh38
rs782581702 Chr.X: 135160448-135160448 on Build GRCh38
rs368825493 Chr.X: 135160373-135160373 on Build GRCh38

View Full Product Details