Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 161364912-161365185 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP126
Entrez Gene ID
257177
Gene Name
cilia and flagella associated protein 126
Gene Aliases
C1orf192, Flattop, Fltp
UniGene
Hs.534593
Gene Chromosome Location
Chr.1: 161364731-161367883 on Build GRCh38
Transcripts
NM_001013625.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3477269 c.511C>G p.P171A Chr.1: 161364988-161364988 on Build GRCh38
5844275 c.509G>A p.G170D Chr.1: 161364990-161364990 on Build GRCh38
6615205 c.362_365delACAG p.D121fs*22 Chr.1: 161365134-161365137 on Build GRCh38
1127296 c.460T>C p.S154P Chr.1: 161365039-161365039 on Build GRCh38
3710372 c.527A>C p.K176T Chr.1: 161364972-161364972 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779645100 Chr.1: 161365013-161365013 on Build GRCh38
rs529245072 Chr.1: 161365040-161365040 on Build GRCh38
rs774572772 Chr.1: 161364941-161364941 on Build GRCh38
rs201764219 Chr.1: 161364995-161364995 on Build GRCh38
rs747758593 Chr.1: 161365068-161365068 on Build GRCh38
rs551537284 Chr.1: 161365009-161365009 on Build GRCh38
rs1051779187 Chr.1: 161365063-161365063 on Build GRCh38
rs945243182 Chr.1: 161364937-161364937 on Build GRCh38
rs776561371 Chr.1: 161365108-161365108 on Build GRCh38
rs764290953 Chr.1: 161364954-161364954 on Build GRCh38
rs750163449 Chr.1: 161364998-161364998 on Build GRCh38
rs901491756 Chr.1: 161365033-161365033 on Build GRCh38
rs751309374 Chr.1: 161365131-161365131 on Build GRCh38
rs16832869 Chr.1: 161364945-161364945 on Build GRCh38
rs757139890 Chr.1: 161364987-161364987 on Build GRCh38
rs997494175 Chr.1: 161364967-161364967 on Build GRCh38
rs764243601 Chr.1: 161365129-161365129 on Build GRCh38
rs267598134 Chr.1: 161365020-161365020 on Build GRCh38
rs571556048 Chr.1: 161365025-161365025 on Build GRCh38
rs754429453 Chr.1: 161365029-161365029 on Build GRCh38
rs569734709 Chr.1: 161365156-161365156 on Build GRCh38
rs771326736 Chr.1: 161365075-161365075 on Build GRCh38
rs532854688 Chr.1: 161365095-161365095 on Build GRCh38
rs1031552137 Chr.1: 161365061-161365061 on Build GRCh38
rs760797668 Chr.1: 161364950-161364950 on Build GRCh38
rs775804555 Chr.1: 161365093-161365093 on Build GRCh38
rs998928106 Chr.1: 161365038-161365038 on Build GRCh38
rs774968603 Chr.1: 161365083-161365083 on Build GRCh38
rs765277909 Chr.1: 161364994-161364994 on Build GRCh38
rs780687466 Chr.1: 161365056-161365056 on Build GRCh38
rs746183973 Chr.1: 161365084-161365084 on Build GRCh38
rs371816162 Chr.1: 161365096-161365096 on Build GRCh38
rs370852007 Chr.1: 161364944-161364944 on Build GRCh38
rs753750882 Chr.1: 161364957-161364957 on Build GRCh38
rs143163178 Chr.1: 161365090-161365090 on Build GRCh38
rs1011405921 Chr.1: 161365137-161365137 on Build GRCh38

View Full Product Details