Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 86567442-86567675 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXC2
Entrez Gene ID
2303
Gene Name
forkhead box C2
Gene Aliases
FKHL14, LD, MFH-1, MFH1
UniGene
Hs.436448
Gene Chromosome Location
Chr.16: 86567251-86568933 on Build GRCh38
Transcripts
NM_005251.2 NR_125795.1

Gene Symbol
FOXC2-AS1
Entrez Gene ID
103752587
Gene Name
FOXC2 antisense RNA 1
Gene Aliases
ODRUL
UniGene
Hs.609366
Gene Chromosome Location
Chr.16: 86565145-86567761 on Build GRCh38
Transcripts
NM_005251.2 NR_125795.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
704501 c.157G>A p.G53S Chr.16: 86567492-86567492 on Build GRCh38
6238040 c.265C>T p.P89S Chr.16: 86567600-86567600 on Build GRCh38
1324234 c.320C>A p.P107H Chr.16: 86567655-86567655 on Build GRCh38
4957732 c.201T>C p.P67P Chr.16: 86567536-86567536 on Build GRCh38
472197 c.128G>A p.G43D Chr.16: 86567463-86567463 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140209790 Chr.16: 86567507-86567507 on Build GRCh38
rs751940772 Chr.16: 86567545-86567545 on Build GRCh38
rs778551960 Chr.16: 86567487-86567487 on Build GRCh38
rs764985786 Chr.16: 86567560-86567560 on Build GRCh38
rs753722640 Chr.16: 86567551-86567551 on Build GRCh38
rs761890099 Chr.16: 86567521-86567521 on Build GRCh38
rs1003983267 Chr.16: 86567533-86567533 on Build GRCh38
rs145253475 Chr.16: 86567518-86567518 on Build GRCh38
rs377509198 Chr.16: 86567627-86567627 on Build GRCh38
rs747725530 Chr.16: 86567489-86567489 on Build GRCh38
rs756695162 Chr.16: 86567593-86567593 on Build GRCh38
rs759417459 Chr.16: 86567617-86567617 on Build GRCh38
rs112024556 Chr.16: 86567506-86567506 on Build GRCh38
rs750865152 Chr.16: 86567586-86567586 on Build GRCh38
rs144326380 Chr.16: 86567610-86567610 on Build GRCh38
rs1007233011 Chr.16: 86567460-86567460 on Build GRCh38
rs886775310 Chr.16: 86567459-86567459 on Build GRCh38
rs957867993 Chr.16: 86567559-86567559 on Build GRCh38
rs770965792 Chr.16: 86567611-86567611 on Build GRCh38
rs369622966 Chr.16: 86567526-86567526 on Build GRCh38
rs779124969 Chr.16: 86567603-86567603 on Build GRCh38
rs778453612 Chr.16: 86567539-86567539 on Build GRCh38
rs926624500 Chr.16: 86567614-86567614 on Build GRCh38
rs556685640 Chr.16: 86567461-86567461 on Build GRCh38
rs143108216 Chr.16: 86567608-86567608 on Build GRCh38
rs746195395 Chr.16: 86567496-86567496 on Build GRCh38
rs752429537 Chr.16: 86567642-86567642 on Build GRCh38
rs757770769 Chr.16: 86567494-86567494 on Build GRCh38
rs758886097 Chr.16: 86567591-86567591 on Build GRCh38
rs764933050 Chr.16: 86567635-86567635 on Build GRCh38
rs920777360 Chr.16: 86567653-86567653 on Build GRCh38
rs749722544 Chr.16: 86567598-86567598 on Build GRCh38
rs773050488 Chr.16: 86567527-86567527 on Build GRCh38
rs371246110 Chr.16: 86567629-86567629 on Build GRCh38
rs104894516 Chr.16: 86567632-86567632 on Build GRCh38
rs768803682 Chr.16: 86567602-86567602 on Build GRCh38
rs1026183412 Chr.16: 86567508-86567508 on Build GRCh38
rs748182598 Chr.16: 86567605-86567605 on Build GRCh38
rs758005564 Chr.16: 86567578-86567578 on Build GRCh38
rs369098719 Chr.16: 86567495-86567495 on Build GRCh38
rs1020830836 Chr.16: 86567592-86567592 on Build GRCh38
rs371100302 Chr.16: 86567620-86567620 on Build GRCh38
rs372781165 Chr.16: 86567476-86567476 on Build GRCh38
rs754671977 Chr.16: 86567554-86567554 on Build GRCh38
rs776500374 Chr.16: 86567613-86567613 on Build GRCh38
rs199862001 Chr.16: 86567468-86567468 on Build GRCh38
rs138472501 Chr.16: 86567566-86567566 on Build GRCh38
rs201393006 Chr.16: 86567469-86567469 on Build GRCh38
rs754723219 Chr.16: 86567483-86567483 on Build GRCh38
rs766061847 Chr.16: 86567536-86567536 on Build GRCh38
rs781696493 Chr.16: 86567581-86567581 on Build GRCh38
rs768926360 Chr.16: 86567512-86567512 on Build GRCh38
rs766012007 Chr.16: 86567470-86567470 on Build GRCh38
rs760703639 Chr.16: 86567528-86567528 on Build GRCh38
rs374000899 Chr.16: 86567594-86567594 on Build GRCh38

View Full Product Details