Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 13405640-13405911 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTBD10
Entrez Gene ID
84280
Gene Name
BTB domain containing 10
Gene Aliases
GMRP-1, GMRP1
UniGene
Hs.332382
Gene Chromosome Location
Chr.11: 13387996-13463297 on Build GRCh38
Transcripts
NM_001297741.1 NM_001297742.1 NM_032320.6 XM_017018405.1 XM_017018406.1 XM_017018407.1 XM_017018408.1 XM_017018409.1 XM_017018410.1 XM_017018411.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1604344 c.999T>G p.Y333* Chr.11: 13405666-13405666 on Build GRCh38
5510494 c.916C>T p.R306W Chr.11: 13405749-13405749 on Build GRCh38
1193669 c.860T>A p.F287Y Chr.11: 13405805-13405805 on Build GRCh38
6217753 c.845G>A p.R282H Chr.11: 13405820-13405820 on Build GRCh38
6068178 c.882C>G p.L294L Chr.11: 13405783-13405783 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs961379323 Chr.11: 13405872-13405872 on Build GRCh38
rs776282519 Chr.11: 13405695-13405695 on Build GRCh38
rs113696304 Chr.11: 13405823-13405823 on Build GRCh38
rs754573161 Chr.11: 13405798-13405798 on Build GRCh38
rs751164213 Chr.11: 13405777-13405777 on Build GRCh38
rs767205028 Chr.11: 13405877-13405877 on Build GRCh38
rs371044722 Chr.11: 13405819-13405819 on Build GRCh38
rs765953914 Chr.11: 13405762-13405762 on Build GRCh38
rs1015803115 Chr.11: 13405829-13405829 on Build GRCh38
rs933764927 Chr.11: 13405852-13405852 on Build GRCh38
rs750405529 Chr.11: 13405750-13405750 on Build GRCh38
rs139063643 Chr.11: 13405738-13405738 on Build GRCh38
rs375035238 Chr.11: 13405820-13405820 on Build GRCh38
rs202219514 Chr.11: 13405682-13405682 on Build GRCh38
rs761668158 Chr.11: 13405734-13405734 on Build GRCh38
rs200389754 Chr.11: 13405753-13405753 on Build GRCh38
rs201139727 Chr.11: 13405717-13405717 on Build GRCh38

View Full Product Details