Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 32238960-32239219 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CMTM8
Entrez Gene ID
152189
Gene Name
CKLF like MARVEL transmembrane domain containing 8
Gene Aliases
CKLFSF8, CKLFSF8-V2
UniGene
Hs.154986
Gene Chromosome Location
Chr.3: 32238679-32370325 on Build GRCh38
Transcripts
NM_001320308.1 NM_178868.4 XM_011533416.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4922473 c.22C>T p.R8C Chr.3: 32238994-32238994 on Build GRCh38
6404533 c.23G>A p.R8H Chr.3: 32238995-32238995 on Build GRCh38
4493723 c.41C>T p.T14I Chr.3: 32239013-32239013 on Build GRCh38
3696032 c.77G>A p.S26N Chr.3: 32239049-32239049 on Build GRCh38
1178639 c.111C>T p.L37L Chr.3: 32239083-32239083 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs529566603 Chr.3: 32239197-32239197 on Build GRCh38
rs780499505 Chr.3: 32239084-32239084 on Build GRCh38
rs939714729 Chr.3: 32239183-32239183 on Build GRCh38
rs775559466 Chr.3: 32239107-32239107 on Build GRCh38
rs779329467 Chr.3: 32239075-32239075 on Build GRCh38
rs146090233 Chr.3: 32239023-32239023 on Build GRCh38
rs531727259 Chr.3: 32239057-32239057 on Build GRCh38
rs375431013 Chr.3: 32239008-32239008 on Build GRCh38
rs779757564 Chr.3: 32239032-32239033 on Build GRCh38
rs746353063 Chr.3: 32239012-32239012 on Build GRCh38
rs546263049 Chr.3: 32239044-32239044 on Build GRCh38
rs768011366 Chr.3: 32239046-32239047 on Build GRCh38
rs775516150 Chr.3: 32239139-32239139 on Build GRCh38
rs752460220 Chr.3: 32238980-32238980 on Build GRCh38
rs754283434 Chr.3: 32238994-32238994 on Build GRCh38
rs772109744 Chr.3: 32239017-32239017 on Build GRCh38
rs372448568 Chr.3: 32239014-32239014 on Build GRCh38
rs753610527 Chr.3: 32239157-32239157 on Build GRCh38
rs371975324 Chr.3: 32239091-32239091 on Build GRCh38
rs898411973 Chr.3: 32239031-32239031 on Build GRCh38
rs765708548 Chr.3: 32239196-32239196 on Build GRCh38
rs767053824 Chr.3: 32238979-32238979 on Build GRCh38
rs770730684 Chr.3: 32239035-32239035 on Build GRCh38
rs772704142 Chr.3: 32239000-32239001 on Build GRCh38
rs761512671 Chr.3: 32239163-32239163 on Build GRCh38
rs774223423 Chr.3: 32239042-32239042 on Build GRCh38
rs376423345 Chr.3: 32239153-32239153 on Build GRCh38
rs780167991 Chr.3: 32239022-32239022 on Build GRCh38
rs76321736 Chr.3: 32239050-32239050 on Build GRCh38
rs553519943 Chr.3: 32238990-32238990 on Build GRCh38
rs748368692 Chr.3: 32239105-32239105 on Build GRCh38
rs763857851 Chr.3: 32239065-32239065 on Build GRCh38
rs376610149 Chr.3: 32239112-32239112 on Build GRCh38
rs772073685 Chr.3: 32239134-32239134 on Build GRCh38
rs953375351 Chr.3: 32239056-32239056 on Build GRCh38
rs753516525 Chr.3: 32239072-32239072 on Build GRCh38
rs761310722 Chr.3: 32239136-32239137 on Build GRCh38
rs945972368 Chr.3: 32239131-32239131 on Build GRCh38
rs769632135 Chr.3: 32239137-32239137 on Build GRCh38
rs757644080 Chr.3: 32238995-32238995 on Build GRCh38
rs762787157 Chr.3: 32239027-32239027 on Build GRCh38
rs569675465 Chr.3: 32239135-32239135 on Build GRCh38
rs750831698 Chr.3: 32239168-32239168 on Build GRCh38
rs148622439 Chr.3: 32238993-32238993 on Build GRCh38
rs770693200 Chr.3: 32239110-32239110 on Build GRCh38
rs72854403 Chr.3: 32239190-32239190 on Build GRCh38
rs764739027 Chr.3: 32239165-32239165 on Build GRCh38
rs892983600 Chr.3: 32239129-32239129 on Build GRCh38
rs944390357 Chr.3: 32239058-32239058 on Build GRCh38
rs369235967 Chr.3: 32239151-32239151 on Build GRCh38
rs781435212 Chr.3: 32239101-32239101 on Build GRCh38
rs772984103 Chr.3: 32239145-32239146 on Build GRCh38
rs760307403 Chr.3: 32239146-32239146 on Build GRCh38
rs776368881 Chr.3: 32239047-32239049 on Build GRCh38
rs757601269 Chr.3: 32239073-32239073 on Build GRCh38
rs750949379 Chr.3: 32239078-32239078 on Build GRCh38
rs758949410 Chr.3: 32239080-32239080 on Build GRCh38
rs746485279 Chr.3: 32239041-32239043 on Build GRCh38
rs747115327 Chr.3: 32239088-32239088 on Build GRCh38
rs776785695 Chr.3: 32239025-32239025 on Build GRCh38
rs894562387 Chr.3: 32239173-32239173 on Build GRCh38
rs200490843 Chr.3: 32239064-32239064 on Build GRCh38
rs1042038128 Chr.3: 32239137-32239137 on Build GRCh38
rs1027099637 Chr.3: 32239191-32239191 on Build GRCh38
rs1056208982 Chr.3: 32238998-32238998 on Build GRCh38
rs1054285017 Chr.3: 32239128-32239128 on Build GRCh38
rs915491324 Chr.3: 32238990-32238993 on Build GRCh38
rs759182915 Chr.3: 32239125-32239125 on Build GRCh38

View Full Product Details