Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 17309554-17309807 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDA1
Entrez Gene ID
79016
Gene Name
DET1 and DDB1 associated 1
Gene Aliases
C19orf58, PCIA1
UniGene
Hs.466154
Gene Chromosome Location
Chr.19: 17309528-17323301 on Build GRCh38
Transcripts
NM_024050.5

SNP Detail

SNP ID Chromosome Location
rs1031582966 Chr.19: 17309781-17309781 on Build GRCh38
rs576237320 Chr.19: 17309639-17309639 on Build GRCh38
rs750156546 Chr.19: 17309610-17309610 on Build GRCh38
rs775378292 Chr.19: 17309670-17309670 on Build GRCh38
rs1006154739 Chr.19: 17309579-17309579 on Build GRCh38
rs780305045 Chr.19: 17309583-17309583 on Build GRCh38
rs774329325 Chr.19: 17309675-17309675 on Build GRCh38
rs971001878 Chr.19: 17309766-17309766 on Build GRCh38
rs370489653 Chr.19: 17309678-17309678 on Build GRCh38
rs766777930 Chr.19: 17309614-17309614 on Build GRCh38
rs201268669 Chr.19: 17309733-17309733 on Build GRCh38
rs1034292839 Chr.19: 17309731-17309731 on Build GRCh38
rs199961906 Chr.19: 17309723-17309723 on Build GRCh38
rs779599481 Chr.19: 17309632-17309632 on Build GRCh38
rs985534727 Chr.19: 17309630-17309630 on Build GRCh38
rs777856884 Chr.19: 17309663-17309663 on Build GRCh38
rs200777492 Chr.19: 17309672-17309672 on Build GRCh38
rs541225874 Chr.19: 17309749-17309749 on Build GRCh38
rs757606453 Chr.19: 17309605-17309610 on Build GRCh38
rs994431406 Chr.19: 17309615-17309615 on Build GRCh38
rs1012598681 Chr.19: 17309734-17309734 on Build GRCh38
rs748220548 Chr.19: 17309633-17309633 on Build GRCh38
rs868052336 Chr.19: 17309626-17309626 on Build GRCh38
rs767062783 Chr.19: 17309604-17309604 on Build GRCh38
rs760394803 Chr.19: 17309682-17309682 on Build GRCh38
rs373202633 Chr.19: 17309625-17309625 on Build GRCh38
rs957548858 Chr.19: 17309786-17309786 on Build GRCh38
rs556175800 Chr.19: 17309585-17309585 on Build GRCh38
rs78265621 Chr.19: 17309687-17309687 on Build GRCh38
rs943462046 Chr.19: 17309598-17309598 on Build GRCh38
rs1038515324 Chr.19: 17309778-17309778 on Build GRCh38
rs35193346 Chr.19: 17309708-17309709 on Build GRCh38
rs1047937410 Chr.19: 17309642-17309642 on Build GRCh38
rs753326442 Chr.19: 17309688-17309688 on Build GRCh38
rs910628226 Chr.19: 17309594-17309594 on Build GRCh38
rs777924018 Chr.19: 17309692-17309692 on Build GRCh38
rs754248077 Chr.19: 17309618-17309618 on Build GRCh38
rs545101186 Chr.19: 17309684-17309684 on Build GRCh38
rs375878246 Chr.19: 17309635-17309635 on Build GRCh38
rs781586252 Chr.19: 17309630-17309641 on Build GRCh38
rs758612261 Chr.19: 17309643-17309643 on Build GRCh38
rs756365744 Chr.19: 17309613-17309613 on Build GRCh38
rs749185153 Chr.19: 17309671-17309671 on Build GRCh38
rs769419845 Chr.19: 17309668-17309668 on Build GRCh38
rs751685410 Chr.19: 17309693-17309693 on Build GRCh38
rs1054733392 Chr.19: 17309592-17309592 on Build GRCh38
rs758345581 Chr.19: 17309690-17309690 on Build GRCh38
rs35729570 Chr.19: 17309620-17309620 on Build GRCh38
rs561286792 Chr.19: 17309775-17309775 on Build GRCh38
rs1040379604 Chr.19: 17309600-17309600 on Build GRCh38
rs1029631746 Chr.19: 17309617-17309617 on Build GRCh38
rs376775549 Chr.19: 17309713-17309713 on Build GRCh38
rs989307558 Chr.19: 17309641-17309641 on Build GRCh38
rs772680962 Chr.19: 17309680-17309680 on Build GRCh38
rs1054862407 Chr.19: 17309750-17309750 on Build GRCh38

View Full Product Details