Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 16033020-16033293 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCC1
Entrez Gene ID
4363
Gene Name
ATP binding cassette subfamily C member 1
Gene Aliases
ABC29, ABCC, GS-X, MRP, MRP1
UniGene
Hs.391464
Gene Chromosome Location
Chr.16: 15949577-16143074 on Build GRCh38
Transcripts
NM_004996.3 XM_011522497.1 XM_011522498.2 XM_017023237.1 XM_017023238.1 XM_017023239.1 XM_017023240.1 XM_017023241.1 XM_017023242.1 XM_017023243.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4394552 c.642C>T p.S214S Chr.16: 16033135-16033135 on Build GRCh38
5698274 c.629A>G p.E210G Chr.16: 16033122-16033122 on Build GRCh38
6590189 c.677+2T>C p.? Chr.16: 16033172-16033172 on Build GRCh38
5698273 c.629A>G p.E210G Chr.16: 16033122-16033122 on Build GRCh38
4578705 c.636C>T p.S212S Chr.16: 16033129-16033129 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766888604 Chr.16: 16033061-16033061 on Build GRCh38
rs202105408 Chr.16: 16033129-16033129 on Build GRCh38
rs867948427 Chr.16: 16033202-16033202 on Build GRCh38
rs762412314 Chr.16: 16033060-16033060 on Build GRCh38
rs775794707 Chr.16: 16033173-16033173 on Build GRCh38
rs760447045 Chr.16: 16033130-16033130 on Build GRCh38
rs774779107 Chr.16: 16033063-16033063 on Build GRCh38
rs575487297 Chr.16: 16033232-16033232 on Build GRCh38
rs8187846 Chr.16: 16033187-16033187 on Build GRCh38
rs901160400 Chr.16: 16033074-16033074 on Build GRCh38
rs1041487976 Chr.16: 16033158-16033158 on Build GRCh38
rs762002192 Chr.16: 16033085-16033085 on Build GRCh38
rs3830390 Chr.16: 16033084-16033085 on Build GRCh38
rs527688744 Chr.16: 16033144-16033144 on Build GRCh38
rs528268276 Chr.16: 16033247-16033247 on Build GRCh38
rs747977371 Chr.16: 16033131-16033131 on Build GRCh38
rs887134343 Chr.16: 16033260-16033260 on Build GRCh38
rs750544452 Chr.16: 16033199-16033199 on Build GRCh38
rs545714984 Chr.16: 16033233-16033233 on Build GRCh38
rs746985426 Chr.16: 16033108-16033108 on Build GRCh38
rs534431004 Chr.16: 16033071-16033071 on Build GRCh38
rs576556933 Chr.16: 16033070-16033070 on Build GRCh38
rs1030063418 Chr.16: 16033240-16033240 on Build GRCh38
rs754912924 Chr.16: 16033201-16033201 on Build GRCh38
rs764352755 Chr.16: 16033184-16033184 on Build GRCh38
rs375353770 Chr.16: 16033137-16033137 on Build GRCh38
rs45492093 Chr.16: 16033187-16033188 on Build GRCh38
rs1039931096 Chr.16: 16033185-16033185 on Build GRCh38
rs756282829 Chr.16: 16033079-16033079 on Build GRCh38
rs563484597 Chr.16: 16033175-16033175 on Build GRCh38
rs1042994509 Chr.16: 16033212-16033219 on Build GRCh38
rs376570605 Chr.16: 16033067-16033067 on Build GRCh38
rs758986221 Chr.16: 16033212-16033215 on Build GRCh38
rs552614067 Chr.16: 16033104-16033104 on Build GRCh38
rs765162708 Chr.16: 16033195-16033195 on Build GRCh38
rs772791393 Chr.16: 16033138-16033138 on Build GRCh38
rs1000375208 Chr.16: 16033183-16033183 on Build GRCh38
rs865872046 Chr.16: 16033103-16033103 on Build GRCh38
rs781307516 Chr.16: 16033215-16033215 on Build GRCh38
rs756842840 Chr.16: 16033165-16033165 on Build GRCh38
rs564253668 Chr.16: 16033241-16033241 on Build GRCh38
rs753921642 Chr.16: 16033083-16033083 on Build GRCh38
rs372049662 Chr.16: 16033097-16033097 on Build GRCh38
rs202023534 Chr.16: 16033139-16033139 on Build GRCh38
rs750999445 Chr.16: 16033211-16033212 on Build GRCh38
rs142564578 Chr.16: 16033181-16033182 on Build GRCh38
rs770415430 Chr.16: 16033140-16033140 on Build GRCh38
rs776788650 Chr.16: 16033071-16033072 on Build GRCh38
rs757350556 Chr.16: 16033189-16033189 on Build GRCh38

View Full Product Details