Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 47723107-47723374 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FNBP4
Entrez Gene ID
23360
Gene Name
formin binding protein 4
Gene Aliases
FBP30
UniGene
Hs.6834
Gene Chromosome Location
Chr.11: 47716510-47767478 on Build GRCh38
Transcripts
NM_001318339.1 NM_015308.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1981950 c.2579C>G p.S860* Chr.11: 47723202-47723202 on Build GRCh38
6284758 c.2524G>A p.V842I Chr.11: 47723257-47723257 on Build GRCh38
3666346 c.2574G>A p.L858L Chr.11: 47723207-47723207 on Build GRCh38
3368380 c.2668C>T p.P890S Chr.11: 47723113-47723113 on Build GRCh38
6315446 c.2474C>A p.A825E Chr.11: 47723307-47723307 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750760023 Chr.11: 47723246-47723247 on Build GRCh38
rs758661397 Chr.11: 47723291-47723291 on Build GRCh38
rs377684522 Chr.11: 47723140-47723140 on Build GRCh38
rs144284794 Chr.11: 47723252-47723252 on Build GRCh38
rs552225968 Chr.11: 47723260-47723260 on Build GRCh38
rs370910213 Chr.11: 47723173-47723173 on Build GRCh38
rs750617792 Chr.11: 47723288-47723288 on Build GRCh38
rs140637382 Chr.11: 47723201-47723201 on Build GRCh38
rs760457742 Chr.11: 47723280-47723280 on Build GRCh38
rs756294204 Chr.11: 47723285-47723305 on Build GRCh38
rs752436326 Chr.11: 47723278-47723278 on Build GRCh38
rs757484594 Chr.11: 47723159-47723159 on Build GRCh38
rs377443770 Chr.11: 47723188-47723188 on Build GRCh38
rs200221203 Chr.11: 47723183-47723183 on Build GRCh38
rs374125956 Chr.11: 47723284-47723284 on Build GRCh38
rs370277154 Chr.11: 47723277-47723277 on Build GRCh38
rs780328249 Chr.11: 47723297-47723297 on Build GRCh38
rs767891410 Chr.11: 47723173-47723190 on Build GRCh38
rs566661800 Chr.11: 47723166-47723166 on Build GRCh38
rs781727247 Chr.11: 47723210-47723210 on Build GRCh38
rs751673974 Chr.11: 47723303-47723303 on Build GRCh38
rs770392191 Chr.11: 47723250-47723250 on Build GRCh38
rs762396248 Chr.11: 47723242-47723242 on Build GRCh38
rs752516715 Chr.11: 47723151-47723151 on Build GRCh38
rs780737126 Chr.11: 47723184-47723184 on Build GRCh38
rs373522050 Chr.11: 47723176-47723176 on Build GRCh38
rs766316813 Chr.11: 47723136-47723136 on Build GRCh38
rs890978097 Chr.11: 47723338-47723338 on Build GRCh38
rs748475626 Chr.11: 47723346-47723346 on Build GRCh38
rs769337975 Chr.11: 47723226-47723226 on Build GRCh38
rs751453260 Chr.11: 47723139-47723139 on Build GRCh38
rs772864117 Chr.11: 47723235-47723235 on Build GRCh38
rs755610762 Chr.11: 47723307-47723307 on Build GRCh38
rs781676627 Chr.11: 47723345-47723345 on Build GRCh38
rs748543947 Chr.11: 47723214-47723214 on Build GRCh38

View Full Product Details