Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 112985816-112986043 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCP2
Entrez Gene ID
167227
Gene Name
decapping mRNA 2
Gene Aliases
NUDT20
UniGene
Hs.443875
Gene Chromosome Location
Chr.5: 112976702-113022195 on Build GRCh38
Transcripts
NM_001242377.1 NM_152624.5 NR_038352.1 XM_017009163.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6047381 c.152T>C p.M51T Chr.5: 112985933-112985933 on Build GRCh38
4185952 c.101G>T p.R34L Chr.5: 112985882-112985882 on Build GRCh38
3776111 c.114G>T p.Q38H Chr.5: 112985895-112985895 on Build GRCh38
3428834 c.92A>G p.N31S Chr.5: 112985873-112985873 on Build GRCh38
3765155 c.185T>A p.I62K Chr.5: 112985966-112985966 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767004909 Chr.5: 112985971-112985972 on Build GRCh38
rs746817650 Chr.5: 112985947-112985947 on Build GRCh38
rs768792813 Chr.5: 112985952-112985952 on Build GRCh38
rs765618193 Chr.5: 112985945-112985945 on Build GRCh38
rs775605039 Chr.5: 112985873-112985873 on Build GRCh38
rs368576692 Chr.5: 112985859-112985859 on Build GRCh38
rs764689436 Chr.5: 112985884-112985884 on Build GRCh38
rs775068122 Chr.5: 112985886-112985886 on Build GRCh38
rs375511134 Chr.5: 112985964-112985964 on Build GRCh38
rs763501540 Chr.5: 112985936-112985938 on Build GRCh38
rs929874682 Chr.5: 112985896-112985896 on Build GRCh38
rs918468082 Chr.5: 112985895-112985895 on Build GRCh38
rs766050029 Chr.5: 112986004-112986004 on Build GRCh38
rs749378408 Chr.5: 112985950-112985950 on Build GRCh38
rs778848312 Chr.5: 112985860-112985860 on Build GRCh38
rs773595507 Chr.5: 112985987-112985987 on Build GRCh38
rs752572068 Chr.5: 112985939-112985939 on Build GRCh38
rs778398996 Chr.5: 112985938-112985938 on Build GRCh38
rs767937195 Chr.5: 112985898-112985898 on Build GRCh38
rs760994227 Chr.5: 112985989-112985989 on Build GRCh38
rs976887250 Chr.5: 112985933-112985933 on Build GRCh38
rs753421028 Chr.5: 112986006-112986006 on Build GRCh38
rs771786420 Chr.5: 112985851-112985851 on Build GRCh38
rs777581644 Chr.5: 112985944-112985944 on Build GRCh38
rs770031556 Chr.5: 112985948-112985948 on Build GRCh38
rs772488929 Chr.5: 112985985-112985985 on Build GRCh38
rs763170777 Chr.5: 112985875-112985875 on Build GRCh38
rs372882501 Chr.5: 112985889-112985889 on Build GRCh38
rs754548516 Chr.5: 112985934-112985934 on Build GRCh38
rs762363805 Chr.5: 112985981-112985981 on Build GRCh38
rs112366745 Chr.5: 112985850-112985850 on Build GRCh38
rs753366850 Chr.5: 112985910-112985910 on Build GRCh38
rs553370185 Chr.5: 112985905-112985905 on Build GRCh38
rs772893862 Chr.5: 112985855-112985855 on Build GRCh38
rs142013240 Chr.5: 112985936-112985936 on Build GRCh38
rs747847949 Chr.5: 112985935-112985935 on Build GRCh38
rs374915154 Chr.5: 112985848-112985848 on Build GRCh38
rs750777134 Chr.5: 112985902-112985902 on Build GRCh38
rs765989351 Chr.5: 112985908-112985908 on Build GRCh38
rs780202226 Chr.5: 112985949-112985949 on Build GRCh38
rs947773456 Chr.5: 112985937-112985937 on Build GRCh38
rs758352574 Chr.5: 112985941-112985941 on Build GRCh38

View Full Product Details